Skip to Content
Merck
All Photos(1)

Key Documents

M4526

Sigma-Aldrich

Minimum Essential Medium Eagle

Alpha Modification, with sodium bicarbonate and Earl's salts, without ʟ-glutamine, ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

αMEM, EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

Product Name

Minimum Essential Medium Eagle, Alpha Modification, with sodium bicarbonate, without L-glutamine, ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

Earle’s salts (5% CO2): yes
sodium pyruvate: yes
glucose: yes
L-glutamine: no
HEPES: no
Hanks’ salts (2% CO2): no
phenol red: yes
NaHCO3: yes
stable glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle (MEM) is a synthetic cell culture media, developed by Harry Eagle. It has higher concentrations of amino acids so the medium more closely approximates the protein composition of cultured mammalian cells. Optional supplementation of non-essential amino acids to the formulations that incorporate either Hanks′ or Earle′s salts has enhanced the uses of this medium. The α modification of MEM with Earle′s balanced salts also known as αMEM, contains non-essential amino acids, sodium pyruvate, and additional vitamins.
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids, some at increased concentrations. In addition, it contains 5 additional vitamins.

Application

Minimum Essential Medium Eagle has been used to maintain:
  • immature cumulus-oocyte complexes from mice
  • human mesenchymal stem cells (MSC)
  • mouse pre-osteoblastic cell line

Reconstitution

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp von Eisenhart-Rothe et al.
Investigative ophthalmology & visual science, 59(12), 5082-5097 (2018-10-30)
Vision loss caused by photoreceptor death represents one of the first symptoms in neuronal ceroid lipofuscinosis, a condition characterized by accumulation of intracellular waste. Cln6nclf mice have a naturally occurring mutation in ceroid-lipofuscinosis neuronal (CLN) protein 6 and are a
Jason M Lajoie et al.
Protein engineering, design & selection : PEDS, 32(5), 219-230 (2019-11-27)
Yeast display immunoprecipitation is a combinatorial library screening platform for the discovery and engineering of antibodies against membrane proteins using detergent-solubilized membrane fractions or cell lysates as antigen sources. Here, we present the extension of this method for the screening
Zhourui Ma et al.
Journal of molecular histology, 51(3), 241-250 (2020-05-14)
Using a large-scale quantitative mesenchymal stem cells (MSCs) membrane proteomics analysis, we identified a large group of ciliary proteins in the MSCs membrane fraction, which prompted us to examine the cilia, intricate organelles that were originally discovered approximately 100 years ago.
Michelle L Churchman et al.
Cancer cell, 33(5), 937-948 (2018-04-24)
Somatic genetic alterations of IKZF1, which encodes the lymphoid transcription factor IKAROS, are common in high-risk B-progenitor acute lymphoblastic leukemia (ALL) and are associated with poor prognosis. Such alterations result in the acquisition of stem cell-like features, overexpression of adhesion
Ricardo Rodrigues-Pinto et al.
Scientific reports, 8(1), 12866-12866 (2018-08-29)
The adult nucleus pulposus originates from the embryonic notochord, but loss of notochordal cells with skeletal maturity in humans is thought to contribute to the onset of intervertebral disc degeneration. Thus, defining the phenotype of human embryonic/fetal notochordal cells is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service