Skip to Content
Merck
All Photos(1)

Key Documents

M0518

Sigma-Aldrich

Minimum Essential Medium Eagle

Joklik Modification, with L-glutamine, without calcium chloride and sodium bicarbonate, suitable for cell culture

Synonym(s):

JMEM, Joklik Modified MEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

HEPES: no
sodium pyruvate: no
L-glutamine: yes
NaHCO3: no
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for suspension cultures.

Quantity

Formulated to contain 11.0 grams of powder per liter of medium.

Reconstitution

Supplement with 2.0 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Rivello et al.
Science advances, 6(40) (2020-10-02)
Despite their important role in metastatic disease, no general method to detect circulating stromal cells (CStCs) exists. Here, we present the Metabolic Assay-Chip (MA-Chip) as a label-free, droplet-based microfluidic approach allowing single-cell extracellular pH measurement for the detection and isolation
Winston T Stauffer et al.
International journal of molecular sciences, 21(4) (2020-02-23)
Activating transcription factor-6 α (ATF6) is one of the three main sensors and effectors of the endoplasmic reticulum (ER) stress response and, as such, it is critical for protecting the heart and other tissues from a variety of environmental insults
Nirmal Verma et al.
Scientific reports, 11(1), 20926-20926 (2021-10-24)
Population-based studies identified an association between a prior pregnancy complicated by gestational diabetes mellitus (GDM) and cardiac hypertrophy and dysfunction later in life. It is however unclear whether GDM initiates this phenotype and what are the underlying mechanisms. We addressed
Giulia Elisa Cambi et al.
Molecular bioSystems, 8(9), 2351-2359 (2012-06-28)
Although angiotensin (Ang)II-induced Janus-activated kinase (JAK)2 phosphorylation was reported to be enhanced in failing human cardiomyocytes, the downstream balance between cardio-protective (signal transducer and activator of transcription-STAT3) and the pro-inflammatory (STAT2 and STAT5) response remains unexplored. Therefore STATs phosphorylation and
Li-Hong Zhang et al.
iScience, 23(3), 100968-100968 (2020-03-22)
R-loops, three-stranded DNA-DNA:RNA hybrid structures, are best known for their deleterious effects on genome stability. The regulatory factors of this fundamental genetic structure remain unclear. Here, we reveal an epigenetic factor that controls R-loop stability. METTL8, a member of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service