M0769
Minimum Essential Medium Eagle
AutoMod™, with Earle′s salts, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Pharma Manufacturing
Synonym(s):
MEM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12171500
NACRES:
NA.75
Recommended Products
Quality Level
form
powder
technique(s)
cell culture | mammalian: suitable
components
NaHCO3: no
phenol red: yes
Earle’s salts (5% CO2): yes
L-glutamine: no
sodium pyruvate: no
HEPES: no
shipped in
ambient
storage temp.
2-8°C
General description
Modification of the original MEM formulation. Contains a succinate buffering system. See formulations table for specific information. Medium may be autoclaved.
Other Notes
Modified for autoclaving.
Quantity
Formulated to contain 9.4 grams of powder per liter of medium.
Reconstitution
Supplement with 0.292 g/L L-glutamine, 2.2 g/L sodium bicarbonate.
Legal Information
Auto-Mod is a trademark of Sigma-Aldrich Co. LLC
supplement
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pasqualino de Antonellis et al.
PloS one, 6(9), e24584-e24584 (2011-09-21)
Through negative regulation of gene expression, microRNAs (miRNAs) can function as oncosuppressors in cancers, and can themselves show altered expression in various tumor types. Here, we have investigated medulloblastoma tumors (MBs), which arise from an early impairment of developmental processes
Simona Giunta et al.
The Journal of cell biology, 190(2), 197-207 (2010-07-28)
The signaling cascade initiated in response to DNA double-strand breaks (DSBs) has been extensively investigated in interphase cells. Here, we show that mitotic cells treated with DSB-inducing agents activate a "primary" DNA damage response (DDR) comprised of early signaling events
Carlos Mendoza-Palomares et al.
Eukaryotic cell, 7(4), 684-697 (2008-02-19)
Cysteine proteases have been shown to be essential virulence factors and drug targets in trypanosomatids and an attractive antidisease vaccine candidate for Trypanosoma congolense. Here, we describe an important amplification of genes encoding cathepsin B-like proteases unique to T. congolense.
Hisashi Miura et al.
Nucleic acids research, 37(3), 778-792 (2008-12-17)
Mammalian tissue- and/or time-specific transcription is primarily regulated in a combinatorial fashion through interactions between a specific set of transcriptional regulatory factors (TRFs) and their cognate cis-regulatory elements located in the regulatory regions. In exploring the DNA regions and TRFs
Michelle Griffin et al.
PloS one, 6(8), e23404-e23404 (2011-08-23)
Non-unions pose complications in fracture management that can be treated using electrical stimulation (ES). Bone marrow mesenchymal stem cells (BMMSCs) are essential in fracture healing; however, the effect of different clinical ES waveforms on BMMSCs cellular activities remains unknown. We
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service