Skip to Content
Merck
All Photos(4)

Key Documents

G7403

Sigma-Aldrich

Glycine

≥99% (titration), suitable for electron microscopy, BioXtra

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Glycine, BioXtra, ≥99% (titration)

product line

BioXtra

Quality Level

Assay

≥99% (titration)

form

powder

technique(s)

electron microscopy: suitable

impurities

Insoluble matter, passes filter test

ign. residue

≤0.05%

loss

≤0.05% loss on drying, HV, 20-26 °C

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

solubility

1 M HCl: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.00001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
NH4+: ≤0.02%
Na: ≤0.02%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.04 at 260 in 1 M HCl at 1 M
≤0.04 at 280 in 1 M HCl at 1 M

SMILES string

NCC(O)=O

UV absorption

λ: 260 nm Amax: ≤0.04
λ: 260 nm Amax: 0.003
λ: 260 nm Amax: 0.006

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Glycine has been used to quench unreacted aldehyde during sample (nuclei) preparation for scanning electron microscope. It has also been used for sample fixation in the process of immunolabeling.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Glycine is a non-essential amino acid. Influx of calcium through the cell membrane is mediated by glycine-gated channel. Glycine participates in the synthesis of porphyrins, purine and serine. It also serves as a competitive agonist for glutamate in binding to the NMDA (N-methyl-D-aspartate) receptors. Glycine synthesis might be increased in rapidly proliferating cancer cells, due to increased demand for the amino acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of the nucleus and nuclear envelope in situ by SEM in tissue culture cells.
Allen TD
Nature Protocols, 2(5), 1180-1184 (2007)
Pre-embedding immunogold labeling to optimize protein localization at subcellular compartments and membrane microdomains of leukocytes.
Melo RC
Nature Protocols, 9(10), 2382-2394 (2014)
Metabolite Profiling Identifies a Key Role for Glycine in Rapid Cancer Cell Proliferation
Mohit Jain
Science, 336(6084), 1040-1044 (2012)
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Bethany L Johnson-Kerner et al.
Human molecular genetics, 24(5), 1420-1431 (2014-11-16)
Giant axonal neuropathy (GAN) is a progressive neurodegenerative disease caused by autosomal recessive mutations in the GAN gene resulting in a loss of a ubiquitously expressed protein, gigaxonin. Gene replacement therapy is a promising strategy for treatment of the disease;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service