Skip to Content
Merck
All Photos(6)

Key Documents

H3375

Sigma-Aldrich

HEPES

HEPES

≥99.5% (titration)

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥99.5% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

cell culture | mammalian: suitable

color

white

pH

5.0-6.5 (25 °C, 238 g/L)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 500 mg/mL, clear, colorless

suitability

suitable for component for culture media
suitable for enzyme extraction

application(s)

diagnostic assay manufacturing
general analytical
life science and biopharma
pharmaceutical

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research.[1] At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.[2]
HEPES, also known as 4-(2-Hydroxyethyl)piperazine-1-ethane-sulfonic acid, is a zwitterionic N-substituted aminosulfonic acid buffer. It is effective in the pH 6.8-8.2 range, with a pKa of 7.48 at 25°C. Recognized as one of the best all-purpose buffers, especially in cell biology, biochemical, and biological research, HEPES is a dipolar ionic buffer and one of the Good′s buffers that does not form significant complexes with most metal ions. This property makes it suitable as a non-coordinating buffer in solutions with metal ions.

Widely utilized in cell culture, HEPES excels at maintaining physiological pH despite fluctuations in carbon dioxide concentration produced by cellular respiration, a feature not as prominent in bicarbonate buffers. It serves as an ampholytic separator for creating pH gradients in isoelectric focusing and exhibits less interference with DNA-restriction enzyme reactions compared to buffers with fewer substituted amine groups, such as Tris. Beyond these applications, HEPES may find application in various biological and biochemical studies, including immunoprecipitation, cell lysis, and live cell imaging. Its versatility makes it an indispensable tool in diverse research applications.

Application

HEPES has been used:
  • As a component in Danieau medium, which is used to prevent melanin pigment formation post gastrulation of zebra-fish embryo[3]
  • As a component of Hanks′ solution used in the degradation testing of Iron[4]
  • As a component of HEPES medium in the transfection of cell culture with plasmids[5]
  • Along with RPMI-1640, L-glutamine, FBS, Sodium pyruvate and β-mercaptoethanol for the culturing of rat insulinoma cells[6]
  • As a supplement in TCM199 medium for washing fresh oocytes required for nuclear transfer studies[7]

Features and Benefits

  • High purity product for biochemical and biological research
  • Suitable for Cell culture
  • Highly soluble in water with a useful pH range of 6.8 - 8.2 and pKa of 7.5 at 25 °C

Other Notes

Go to Redi-Dri Product Listing RDD002
Easily compare specifications for HEPES products with the HEPES specification table.
For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caveolae are highly immobile plasma membrane microdomains, which are not involved in constitutive endocytic trafficking.
Thomsen P, et al.
Molecular Biology of the Cell, 13(1), 238-250 (2002)
Electroformed pure iron as a new biomaterial for degradable stents: In vitro degradation and preliminary cell viability studies.
Moravej M, et al.
Acta Biomaterialia, 6(5), 1843-1851 (2010)
Culturing INS-1 cells on CDPGYIGSR-, RGD-and fibronectin surfaces improves insulin secretion and cell proliferation.
Kuehn C, et al.
Acta Biomaterialia, 8(2), 619-626 (2012)
High-resolution in situ hybridization to whole-mount zebrafish embryos.
Thisse C and Thisse B
Nature Protocols, 3(1), 59-69 (2008)
Structural, metabolic and developmental evaluation of ovulated rabbit oocytes before and after cryopreservation by vitrification and slow freezing.
Salvetti P, et al.
Theriogenology, 74(5), 847-855 (2010)

Questions

1–6 of 6 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What concentration of HEPES should be used for cell culture without CO2, to maintain optimal pH?

    1 answer
    1. This product has not been tested for cell culture use. A better choice is H4034 HEPES BioPerformance Certified, ?99.5% (titration), suitable for cell culture, or one of the other products referred to in the Technical Bulletin found in the documentation section of the product page:
      https://www.sigmaaldrich.com/product/sigma/h4034#product-documentation

      Helpful?

  4. What is the concentration of HEPES used in cell culture?

    1 answer
    1. HEPES is typically used in cell culture in the range of 10-25 mM.

      Helpful?

  5. What is the purpose of using HEPES in cell culture?

    1 answer
    1. HEPES provides supplemental buffering for media in pH range of 7.2 to 7.6. It has no nutritional value to cells.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

Active Filters

  1. Southampton
    • Review 1
    • Vote 1
    5 out of 5 stars.

    Arrived quickly and well packaged

    Arrived very quickly and well packaged, thank you.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service