Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1323-01

Glycine

Cytiva 17-1323-01, pack of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352209
NACRES:
NA.56

form

solid

feature

wetted part: no

packaging

pack of 500 g

manufacturer/tradename

Cytiva 17-1323-01

color

White

solubility

acetone: soluble (Slightly soluble in acetone)
cold water: soluble (Soluble in cold water and hot water)
diethyl ether: soluble (Insoluble in diethyl ether)

Related Categories

General description

Glycine is frequently used as a component of electrophoresis buffers, especially in the Laemmli discontinuous buffer system for SDS-PAGE.

Features and Benefits

  • Highly pure PlusOne Glycine made by Cytiva.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 5 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service