Skip to Content
Merck
All Photos(1)

Key Documents

67419

Sigma-Aldrich

Glycine 1 M solution

Synonym(s):

Glycine solution, Additive Screening Solution 47/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

form

liquid

Quality Level

density

1.03 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[N+H3]CC(=O)[O-]

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycine functions as an allosteric modulator for glutamate. It is implicated in the biosynthesis of porphyrins and purines. Glycine acts as a neurotransmitter in the central nervous system.
Glycine is the simplest polar uncharged amino acid, containing a single hydrogen group for an R group. Glycine works as a buffer or stabilizes the pH of substances with alkaline or acidic nature.
Glycine 1M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.

Application

Glycine 1 M solution has been used as a component in culture medium to prepare cells for electroporation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haschek and Rousseaux's Handbook of Toxicologic Pathology (2013)
Essentials of Medical Biochemistry: With Clinical Cases (2011)
Bacterial Adaptation to the Host's Diet Is a Key Evolutionary Force Shaping Drosophila-Lactobacillus Symbiosis
Martino ME, et al.
Cell host & microbe, 24(1), 109-119 (2018)
Nutritional and Herbal Therapies for Children and Adolescents: A Handbook for Mental Health Clinicians, 24(1), 109-119 (2010)
United States International Trade Commission
Glycine from the People's Republic of China (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service