Skip to Content
Merck
All Photos(3)

Documents

E4637

Sigma-Aldrich

Elaidic acid

≥99.0% (GC)

Synonym(s):

Oleic acid, trans-9-Octadecenoic acid, trans-Oleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726543
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Assay

≥99.0% (GC)

form

powder

bp

288 °C/100 mmHg (lit.)

mp

42-44 °C (lit.)

functional group

oleic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+

InChI key

ZQPPMHVWECSIRJ-MDZDMXLPSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U McCloy et al.
Journal of lipid research, 45(3), 474-485 (2003-12-18)
Altered use of different dietary fatty acids may contribute to several chronic diseases, including obesity, noninsulin-dependent diabetes mellitus, and cardiovascular disease. However, few comparative data are available to support this link, so the goal of the present study was to
Nima Sanadgol et al.
Clinical biochemistry, 43(12), 968-972 (2010-05-11)
Elaidic acid, the predominant trans-fatty acid in industrially hydrogenated oils, exists on high levels in Iranian hydrogenated oils and margarines. This study was undertaken to investigate the effect of elaidic acid and its cis-counterpart oleic acid on expression of ICAM-1
G Andrei et al.
Antiviral research, 45(3), 157-167 (2000-04-20)
A fatty acid derivative of ganciclovir (GCV), elaidic acid ganciclovir (E-GCV), has been evaluated for its inhibitory activity against laboratory and clinical strains of herpes simplex type 1 (HSV-1) and type 2 (HSV-2), varicella-zoster virus (VZV) and human cytomegalovirus (HCMV)
Patricia Burriel et al.
Physical review letters, 100(13), 134503-134503 (2008-06-04)
An anomalously long transient is needed to achieve a steady pressurization of a fluid when forced to flow through micronarrowed channels under constant mechanical driving. This phenomenon, known as the "bottleneck effect" is here revisited from a different perspective, by
Suzanne E Dorfman et al.
Obesity (Silver Spring, Md.), 17(6), 1200-1207 (2009-07-09)
Dietary trans-fatty acids are associated with increased risk of cardiovascular disease and have been implicated in the incidence of obesity and type 2 diabetes mellitus (T2DM). It is established that high-fat saturated diets, relative to low-fat diets, induce adiposity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service