Skip to Content
Merck

Skip To

E3385

Erucic acid

≥99% (capillary GC)

Synonym(s):

cis-Erucic acid, cis-13-Docosenoic acid, Prifac 2990

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)11COOH
CAS Number:
Molecular Weight:
338.57
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
204-011-3
Beilstein/REAXYS Number:
1728049
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Level

assay

≥99% (capillary GC)

form

powder

bp

358 °C/400 mmHg (lit.)

mp

28-32 °C (lit.)

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCCC(O)=O

InChI

1S/C22H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H,23,24)/b10-9-

InChI key

DPUOLQHDNGRHBS-KTKRTIGZSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
45630N1514T6543
assay

≥99% (capillary GC)

assay

~90% (GC)

assay

≥99% (capillary GC)

assay

≥99% (capillary GC)

biological source

synthetic (organic)

biological source

-

biological source

plant

biological source

synthetic (organic)

functional group

carboxylic acid

functional group

carboxylic acid

functional group

carboxylic acid

functional group

carboxylic acid

shipped in

ambient

shipped in

-

shipped in

ambient

shipped in

ambient

form

powder

form

powder or crystals

form

powder

form

powder

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Erucic acid


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Arun Jagannath et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 122(6), 1091-1103 (2010-12-29)
Oil content and oil quality fractions (viz., oleic, linoleic and linolenic acid) are strongly influenced by the erucic acid pathway in oilseed Brassicas. Low levels of erucic acid in seed oil increases oleic acid content to nutritionally desirable levels, but
Shiva Shanker Kaki et al.
Biotechnology and bioengineering, 110(1), 78-86 (2012-07-20)
The enzymatic conversion of mixtures of multiple substrates was studied quantitatively, based on established methodology used for the enzymatic kinetic resolution of racemic mixtures, involving the use of competitive factors: ratios of specificity constants (k(cat)/K(M)) of substrate pairs. The competitive
Gang Wu et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 116(4), 491-499 (2007-12-14)
The fatty acid elongase 1 (FAE1) gene is a key gene in the erucic acid biosynthesis in rapeseed. The complete coding sequences of the FAE1 gene were isolated separately from eight high and zero erucic acid rapeseed cultivars (Brassica napus



Global Trade Item Number

SKUGTIN
E3385-5G04061833057520
E3385-1G04061833603260

Questions

Reviews

No rating value

Active Filters