Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
L1376
Linoleic acid
≥98%
Synonym(s):
α-Linoleic acid, cis-9,cis-12-Octadecadienoic acid
About This Item
Recommended Products
biological source
plant oil (safflower)
Quality Level
Assay
≥98%
form
liquid
color
colorless
refractive index
n20/D 1.466 (lit.)
bp
229-230 °C/16 mmHg (lit.)
mp
−5 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
application(s)
life science and biopharma
functional group
carboxylic acid
lipid type
omega FAs
shipped in
ambient
storage temp.
−20°C
SMILES string
OC(CCCCCCC/C=C\C/C=C\CCCCC)=O
InChI
1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-
InChI key
OYHQOLUKZRVURQ-HZJYTTRNSA-N
Gene Information
human ... SOAT1(6646) , SOAT2(8435)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
I noticed it comes as a liquid. What does it coke dissolved in?
1 answer-
This product is not diluted with any solvent. The physical form is a liquid. It is considered a 'neat' product with a minimum purity of 99%. The density is 0.902 g/mL at room temperature. The product is sold by weight, the 5 gram package would be approx. 5.5 mL.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service