Skip to Content
Merck
All Photos(5)

Key Documents

D1313

Sigma-Aldrich

JumpStart REDAccuTaq® LA DNA Polymerase

Long and accurate hot-start Taq with inert dye, 10X buffer included

Synonym(s):

High fidelity Taq, Hot start DNA polymerase, Hot start Taq, Long Taq

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106314
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 250 reactions
sufficient for 50 reactions

feature

Long & Accurate PCR
dNTPs included: no
hotstart

concentration

1 unit/μL

technique(s)

PCR: suitable

color

red

input

purified DNA

suitability

suitable for PCR

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart REDAccuTaq® LA DNA Polymerase contains AccuTaq long and accurate (LA) DNA polymerase, an inert red dye, and JumpStart Taq antibody. This enzyme is suitable for long-distance and high-fidelity PCR, multiplex PCR, and PCR amplification of targets with variable lengths, such as amplification of cDNA libraries. It enables the amplification from 0.25 to 22 kb for complex genomic DNA and up to 40 kb for less complex templates. JumpStart Taq antibody is designed to minimize non-specific amplification while increasing target yield. Unlike other Hot-start methods (i.e. chemical inactivation), the JumpStart Taq antibody does not require a pre-incubation step before cycling because polymerase activity is fully restored during the first denaturation cycle of the PCR reaction. The PCR product can be easily separated from the red dye by standard purification methods. The inert red dye has does not affect automated sequencing, restriction enzyme digestion, ligation, or other downstream applications. Its high fidelity makes it the enzyme of choice when performing amplifications where a low error frequency is critical, such as in RT-PCR and cloning.

Application

JumpStart REDAccuTaq LA DNA Polymerase is a unique enzyme blend that is capable of generating long PCR fragments, from 0.25 kb to 40 kb, with high fidelity, increased specificity and yield. JumpStart REDAccuTaq DNA polymerase combines Sigma′s AccuTaq LA DNA polymerase and JumpStart Taq antibody with an inert red dye. This specially formulated hot start enzyme mix achieves greater yields, enhances sensitivity and results in higher fidelity (6.5×) in comparison to standard Taq or other Long and Accurate enzyme blends. Its high fidelity makes it the enzyme of choice when performing amplifications where a low error frequency is critical, such as in RT-PCR and cloning.

JumpStart REDAccuTaq LA DNA Polymerase also contains the hot start mechanism of the JumpStart Taq antibody. JumpStart Taq antibody is designed to minimize non-specific amplification while increasing target yield. Unlike other hot-start methods (i.e. chemical inactivation), JumpStart Taq antibody does not require a pre-incubation step prior to cycling because polymerase activity is fully restored during the first denaturation cycle of the PCR reaction.

The inert red dye provides quick recognition and confirmation of appropriate mixing. An aliquot of the samples (5-10 μL) may be loaded directly onto an agarose gel following PCR. The red dye migrates slightly faster than bromophenol blue at the same rate as a 125 base pair fragment.

The PCR product can be easily separated from the dye by standard purification methods. The inert red dye has does not effect automated sequencing, restriction enzyme digestion, ligation or other downstream applications.
JumpStart REDAccuTaq® LA DNA Polymerase has been used for amplifying genomic DNA. It has also been used in polymerase chain reaction (PCR) for gene cloning.

Features and Benefits

  • JumpStart REDAccuTaq LA DNA polymerase, an antibody inactivated hot start enzyme, is designed to minimize non-specific amplification while increasing target yield & specificity
  • Up to 6.5X greater fidelity in comparison to Taq DNA polymerase making it the ideal enzyme for multiplex PCR
  • Produce amplicons up to 22 kb with genomic templates and up to 40 kb with less complex templates such as lambda or bacterial genomic DNA
  • Reduce set-up time and eliminate concerns associated with manual or wax Hot Start methods
  • Dye allows for quick visual confirmation that reagent has been added and mixed properly
  • Direct loading onto an agarose gel without additional dyes

Packaging

Supplied with optimized 10× reaction buffer

Unit Definition

One unit incorporates 10 nmol of total dNTPs into acid precipitable DNA in 30 min at 74 °C.

Other Notes

View more detailed information on JumpStart REDTaq and Accutaq enzymes at www.sigma-aldrich.com/hotstart.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
JumpStart is a trademark of Sigma-Aldrich Co. LLC
REDAccuTaq is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas M Sesterhenn et al.
Molecular genetics and genomics : MGG, 283(1), 63-72 (2009-11-19)
With the exception of a few genes, most of the mitochondrial (mt) genome of Pneumocystis carinii has not previously been sequenced. Shotgun sequences generated as a result of the Pneumocystis Genome Project (PGP) were assembled with the gap4 assembly program
Single loci detection and karyotyping using small target FISH on maize somatic chromosomes
Lamb J C, et al.
Genetics (2007)
Mark Sharkey et al.
Journal of virology, 79(8), 5203-5210 (2005-03-30)
Current regimens for the management of human immunodeficiency virus type 1 (HIV-1) infection suppress plasma viremia to below detectable levels for prolonged intervals. Nevertheless, there is a rapid resumption in plasma viremia if therapy is interrupted. Attempts to characterize the
Roberto Giorda et al.
American journal of human genetics, 85(3), 394-400 (2009-09-01)
Submicroscopic copy-number variations make a considerable contribution to the genetic etiology of human disease. We have analyzed subjects with idiopathic mental retardation (MR) by using whole-genome oligonucleotide-based array comparative genomic hybridization (aCGH) and identified familial and de novo recurrent Xp11.22-p11.23
Scott T Lefurgy et al.
Protein science : a publication of the Protein Society, 16(12), 2636-2646 (2007-11-22)
In class C beta-lactamases, the strictly conserved Asn152 forms part of an extended active-site hydrogen-bonding network. To probe its role in catalysis, all 19 mutants of Enterobacter cloacae P99 cephalosporinase Asn152 were simultaneously constructed and screened in Escherichia coli for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service