Skip to Content
Merck
All Photos(2)

Documents

C1889

Sigma-Aldrich

Collagenase from Clostridium histolyticum

0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Quality Level

sterility

0.2 μm filtered

type

Type IV-S

form

powder

specific activity

≥125 CDU/mg solid
0.5-5.0 FALGPA units/mg solid

mol wt

68-130 kDa

suitability

suitable for release of physiologically active rat hepatocytes

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Clostridium histolyticum consists of two classes of collagenases, which cleaves at multiple cleavage sites within the collagen triple helix. It is more effective than mammalian collagenases. Clostridial collagenase is implicated in bacterial invasion in gas gangrene.

Application

Collagenase from Clostridium histolyticum has been used to dissociate tissue samples such as spleens and testicular tissue.
Collagenase has been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products (AGE). The enzyme has also been used along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, lung epithelium and many other tissues. It is also effective in liver and kidney perfusion studies, digestion of pancreas, and isolation of nonparenchymal hepatocytes. When this enzyme is tested for suitability for the release of hepatocytes. The collagenase is used at approximately 1 mg/mL in a total volume of 100 mL for each rat liver.

Biochem/physiol Actions

Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa (pH optimum: 6.3-8.8). The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL is used for digestion. The duration of reaction varies from 15 minutes to several hours, which enables efficient cell dissociation without causing too much cell death. Krebs Ringer buffer with calcium and BSA is preffered and Zn2+ is required for its activity.
Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.

Caution

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

Prepared from Type IV (C5138). A stock solution may be prepared by dissolving 0.05-0.1 mg/mL of collagenase in 50 mM TES buffer containing 0.36 mM calcium chloride (TESCA) (pH 7.4) at 37 °C. This product also contains clostripain, nonspecific neutral protease, and tryptic activities.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial artificial chromosomes (BACs)-on-Beads? as a diagnostic platform for the rapid aneuploidy screening of products of conception
Sheath KL, et al.
Molecular Medicine Reports, 8(2), 650-654 (2013)
Ting Liu et al.
Gut, 70(10), 1965-1977 (2020-12-03)
It remains controversial whether tumour mutational burden (TMB) or neoantigens are prognostic markers in hepatocellular carcinoma (HCC). This study aimed to define the function of TMB or neoantigens in antitumour immunotherapy. Neoantigens of patients (n=56) were analysed by pVAC tools
Rebecca M Reese et al.
Scientific reports, 10(1), 979-979 (2020-01-24)
The transient receptor potential (TRP) superfamily of ion channels has garnered significant attention by the pharmaceutical industry. In particular, TRP channels showing high levels of expression in sensory neurons such as TRPV1, TRPA1, and TRPM8, have been considered as targets
Generation of Mouse Spermatogonial Stem-Cell-Colonies in A Non-Adherent Culture
Azizi H, et al.
Cell Journal, 19(2), 238-238 (2017)
V Secq et al.
Cell death & disease, 6, e1592-e1592 (2015-01-16)
Pancreatic ductal adenocarcinoma (PDA) is a critical health issue in the field of cancer, with few therapeutic options. Evidence supports an implication of the intratumoral microenvironment (stroma) on PDA progression. However, its contribution to the role of neuroplastic changes within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service