Skip to Content
Merck

C0773

Collagenase from Clostridium histolyticum

high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)

Synonym(s):

Clostridiopeptidase A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-582-9
MDL number:
EC Number:
Specific activity:
≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

type

Type VII

form

lyophilized powder

specific activity

≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)

mol wt

68-130 kDa

purified by

chromatography

composition

Protein, ≥85% biuret

solubility

TESCA buffer (50 mM TES, 0.36 mM Calcium chloride, pH 7.4): soluble 0.05-0.1 mg/mL at 37 °C

foreign activity

caseinase and clostripain ≤1 units/mg protein

storage temp.

−20°C

Application

Collagenase has been used in the preparation of arterial tissue for the study of advanced glycosylation end products (AGE). The enzyme has also been used along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, lung epithelium and many other tissues. It is also effective in liver and kidney perfusion studies, digestion of pancreas, and isolation of nonparenchymal hepatocytes.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. Optimal pH ranges from 6.3-8.8. The enzyme recognizes the sequence -R-Pro-8-X-Gly-Pro-R-, where X is most often a neutral amino acid.

Physical form

Lyophilized powder containing calcium chloride

Preparation Note

A stock solution may be prepared by dissolving 0.05 - 0.1 mg of collagenase in one mL of 50 mM TES buffer containing 0.36 mM calcium chloride (TESCA), pH 7.4 at 37 °C. Use cold buffer.
Please note the material will not dissolve completely and few sediments may be seen. The sediments do not affect the activity of the solution.

Other Notes

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
C0773-15KU04061833452073
C0773-60KU04061832699844
C0773-30KU04061832699820
C0773-1.5KU04061833452066
C0773-3KU04061832699837
C0773-7.5KU04061832699851