Skip to Content
Merck
All Photos(1)

Key Documents

SCR103

Sigma-Aldrich

Collagenase type I

The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.

Synonym(s):

Collagenase Type I from Clostridium histolyticum, Tissue dissociation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

powder

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)

shipped in

wet ice

General description

Collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means. The preparation also may contain caseinase, clostripain, and tryptic activities. This preparation can be used to dissociate cells in culture, and has been successfully used to passage human embryonic stem cells cultured with Millipore’s HEScGROTM Animal Component-Free Medium.

Packaging

1 vial containing 250mg of lyophilized powder

Quality

Each lot of collagenase type I is assayed via a modification of Mandl (1964), with the extent of collagen breakdown determined using the colorimetric ninhydrin method of Moore and Stein (1948). Each lot is then tested for successful passage of human embryonic stem cells cultured in Millipore′s HEScGRO Medium for hES cells. Cells are characterized for correct marker expression and morphology after five passages.

Physical form

Collagenase type I is supplied as a dialyzed, lyophilized powder.

Storage and Stability

Collagenase type I should be stored in its powdered form at 2-8oC until ready to use. Solutions of collagenase type I can be aliquotted and stored at -20oC, but should not be repeatedly thawed and re-frozen.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu-Lu Li et al.
Frontiers in cellular and infection microbiology, 10, 581535-581535 (2021-01-26)
Pseudomonas aeruginosa (PA) is one of the most common Gram-negative bacteria causing hospital-acquired pulmonary infection, with high drug resistance and mortality. Therefore, it is urgent to introduce new non-antibiotic treatment strategies. Mesenchymal stem cells (MSCs), as important members of the
Nigel S Michki et al.
Cell reports, 35(4), 109039-109039 (2021-04-29)
The Drosophila type II neuroblast lineages present an attractive model to investigate the neurogenesis and differentiation process as they adapt to a process similar to that in the human outer subventricular zone. We perform targeted single-cell mRNA sequencing in third
Yiya Yang et al.
International urology and nephrology, 53(6), 1247-1254 (2021-05-05)
High glucose (HG) induces the production of transforming growth factor (TGF)-β and reactive oxygen species, which further activates JAK/STAT signaling and promotes the synthesis of matrix proteins, contributes to the pathophysiological processes of diabetic nephropathy. This study aims to investigate
Jinfei Fu et al.
American journal of cancer research, 10(12), 4234-4250 (2021-01-09)
Cytotoxic CD8+ T cell exhaustion is one of the mechanisms underlying the tumor immune escape. The paradigm-shifting immune checkpoint therapy can mitigate CD8+ T lymphocyte exhaustion, reinvigorate the anticancer immunity, and achieve durable tumor regression for some patients. Emerging evidence
Jun Zeng et al.
Respiratory research, 21(1), 328-328 (2020-12-23)
Thymosin β10 (TMSB10) has been reported to play a protumorigenic role in a majority of solid cancers. However, the existence of TMSB10 in immune microenvironment may contribute to the pathogenesis of lung adenocarcinoma has not been previously explored. TAMs-associated TMSB10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service