Skip to Content
Merck
All Photos(1)

Key Documents

C2-BIOC

Sigma-Aldrich

Collagenase Type II, Cls II

Synonym(s):

Collagen A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.78
Pricing and availability is not currently available.

General description

Collagenase Type II (Cls II) is one of the main tissue dissociation enzymes produced by Clostridium histolyticum. It aids to digest the smaller collagen fragments produced by Type I collagenase. Collagenases, a member of the matrix metalloproteinase (MMPs) family, digests collagen in the extracellular matrix (ECM). Collagens are triple helix structural proteins comprising three collagen polypeptides with high tensile strength.[1]
This product is identical to Biochrom GmbH part numbers C2-22 and C2-28. Enzyme blending from collagenase, clostripain, with tryptic and proteolytic activities from Clostridium histolyticum. This preparation shows a high clostripain activity with the tryptic activity close to type I. Suitable for liver, bone, thyroid, heart, and salivary gland tissue. Specific activity is 125 to 250 Mandl units per milligram of powdered substance.

Application

Collagenase Type II (Cls II) has been used:
  • as a component of Roswell park memorial Institute (RPMI)-1640 media to digest hepatic tissues[2]
  • to perfuse murine heart for the isolation of epicardial stromal cells (EpiSC)[3]
  • to digest collagenase and inject it into the intrarenal aorta abdominalis to perfuse the kidneys[4]


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Yao Zhang et al.
Stem cell research & therapy, 13(1), 340-340 (2022-07-27)
Nucleus pulposus mesenchymal stem cells (NPMSCs) transplantation is a promising treatment for intervertebral disc degeneration (IVDD). However, the transplanted NPMSCs exhibited weak cell proliferation, high cell apoptosis, and a low ability to resist the harsh microenvironment of the degenerated intervertebral
Fanglong Song et al.
Inflammation, 46(2), 739-751 (2022-12-09)
Osteoarthritis (OA) is believed to be linked with cartilage degeneration, subchondral bone sclerosis, and synovial inflammation that lead to joint failure, and yet treatment that can effectively reverse the pathological process of the disease still not exists. Recent evidence suggests
Tissue dissociation enzymes for adipose stromal vascular fraction cell isolation: a review
Lockhart RA, et al.
Journal of stem cell research & therapy (2015)
Yuan Yang et al.
Toxicology and applied pharmacology, 410, 115364-115364 (2020-12-09)
Semaphorin (Sema) 3A and Sema 4A are immunomodulatory molecules with a common receptor, neuropilin-1 (NRP-1), on the immune cells. Sema 3A binds to NRP-1 and inhibits T cell activation and inflammation, while Sema 4A binds to NRP-1 and promotes T
Pei-Long Liu et al.
Analytical cellular pathology (Amsterdam), 2023, 7573165-7573165 (2023-05-18)
Diabetic neuropathic osteoarthropathy (DNOAP) is a rare and easily missed complication for diabetes that leads to increased morbidity and mortality. DNOAP is characterized by progressive destruction of bone and joint, but its pathogenesis remains elusive. We herein aimed to investigate

Questions

1–5 of 5 Questions  
  1. Hello Can I use the collagenase type II to digest human adipose tissue and isolate mesenchymal stem cells? Thank you

    1 answer
    1. This product has not been tested for the application of digesting human adipose tissue to isolate mesenchymal stem cells. Testing the product empirically is necessary.

      Helpful?

  2. Dear Sir, I stored reconstituted collagenase in a -20 degrees fridge after diluting and using some of it, after 4months is the enzyme still viable for use? It was left in -20 degrees all the while, thanks

    1 answer
    1. The solution stability of this product has not been determined. However, other collagenase products available have been determined to be stable when stored in solution at –20°C. Repeated freeze/thaw cycles are not recommended and may result in a decline in activity. Please see the link below to review the product datasheet for C2139, a collagenase preparation for the isolation of adipocytes and hepatocytes:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/173/127/c2139pis.pdf

      Helpful?

  3. Kann die Kollagenase für die enzymatische Verdauung und Isolierung von Hühnerembryoosteoblasten verwendet werden? Wie ist die Reinheit des Produkts?

    1 answer
    1. This product has not been tested for use in isolating osteoblasts from chick embryos. Currently, there is no collagenase product option that has been tested for suitability in this application. The product is generally used for bone tissue digests. However the end user would have to determine suitability experimentally.

      A preliminary search of PubMed and Google Scholar did not find research articles citing this specific application.  The working enzyme concentration range is typically 0.1 to 0.2% (w/v), depending on the delicate nature of the tissue substrate. This product is tested for enzymatic activity. The analytical purity has not been determined.

      Helpful?

  4. 골관절염 모델링에 있어 collagenase type 2 를 500 U로 Intra-articular injection 해야합니다만, spec sheet 에 쓰여있는 바로는 units/mg Solid (Collagen) 이 >= 125 로 되어있는데 mg 당 정확한 unit을 알수는 없나요 ?

    1 answer
    1. The exact value is reported on the lot specific Certificate of Analysis. Please see the link below to access a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sial/c2bioc#product-documentation

      Helpful?

  5. Dear sir I was bought collagenase before, but I can't find my instruction manual. Can I have a copy? I need to prepare 0.2% solution to dissolve tumor cells, and I want to grow solute for greenhouse configuration and two for dissolving cells.

    1 answer
    1. Unfortunately, no user manual is provided with this product. It is recommended to prepare working solution by dissolving the enzyme in water to a final concentration of 100mg/mL. Dilute with PBS (Ca2+/Mg2+-free) to a final concentration of 1mg/mL and filter through a sterile 0.2μm membrane filter.

      Another Collagenase Type II product under the Sigma-Aldrich brand, C6885, does offer a product information sheet. While the lot-specific activity between the two products varies, the storage, handling, and functional information would be the same. See the link below to review this document:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/321/500/c6885pis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service