Skip to Content
Merck
All Photos(2)

Key Documents

C9697

Sigma-Aldrich

Collagenase from Clostridium histolyticum

lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Clostridium histolyticum

Quality Level

sterility

0.2 μm filtered

form

lyophilized powder (from 0.2 μm filtered solution)

specific activity

≥800 units/mg solid

mol wt

68-130 kDa

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

pH

7.4

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Collagenase is a protease which cleaves the triple-helical protein called collagen. There are three types of tissue collagenases, and these belong to the matrix metalloproteinases (MMP) family.[1] Collagenase obtained from Clostridium histolyticum has a very strong activity, as it digests collagen from both ends, at temperatures as low as 4-10°C.[2] Crude collagenase mixtures contain two major enzyme types namely, collagenase and clostripain.[3]

Application

Collagenase from Clostridium histolyticum is used for the following applications:

  • Sertoli cell isolation[4]
  • Used in the comparison of enzymatic methods[5]
  • Tissue preparations for immunocytochemistry[6]
  • Testicular sperm extraction[7]
  • Preparation of single cell suspensions[8]
  • Immunofluorescence[9]

This product is suitable for the disaggregation of human tumor, mouse kidney, human adult and fetal brain, lung and many other epithelia tissues. It has also been shown to be effective in liver and kidney perfusion studies, digestion of pancreas and hepatocyte preparation. Collagenase has also been used in the preparation of arterial tissue for the study of Advanced Glycosylation End Products. This enzyme has been tested for the release of heptatocytes at a concentration of approximately 1mg/mL. Concentrations for digestion range from 0.1 to 5mg/mL.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular materialThe action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

Caution

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

Solutions are prepared from type XI at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37°C).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyssa A Leystra et al.
Cancer reports (Hoboken, N.J.), e1459-e1459 (2021-07-11)
Data are steadily accruing that demonstrate that intestinal tumors are frequently derived from multiple founding cells, resulting in tumors comprised of distinct ancestral clones that might cooperate or alternatively compete, thereby potentially impacting different phases of the disease process. We
Katharine Striedinger et al.
STAR protocols, 2(1), 100302-100302 (2021-02-09)
Regeneration and repair of skeletal muscle is driven by tissue-specific progenitor cells called satellite cells, which occupy a minority of the cells in the muscle. This protocol provides researchers with techniques to efficiently isolate and purify functional satellite cells from
Rosa Scala et al.
Frontiers in pharmacology, 11, 604885-604885 (2020-12-18)
Cantù syndrome (CS) arises from mutations in ABCC9 and KCNJ8 genes that lead to gain of function (GOF) of ATP-sensitive potassium (KATP) channels containing SUR2A and Kir6.1 subunits, respectively, of KATP channels. Pathological consequences of CS have been reported for
Christopher Chase Bolt et al.
Nature communications, 12(1), 5013-5013 (2021-08-20)
Human families with chromosomal rearrangements at 2q31, where the human HOXD locus maps, display mesomelic dysplasia, a severe shortening and bending of the limb. In mice, the dominant Ulnaless inversion of the HoxD cluster produces a similar phenotype suggesting the
E Crabbé et al.
Human reproduction (Oxford, England), 12(8), 1682-1687 (1997-08-01)
Intracytoplasmic sperm injection (ICSI) with testicular spermatozoa and ejaculated spermatids has been applied successfully to treat infertility because of azoospermia due to obstruction or testicular failure. In most cases, free spermatozoa are recovered from testicular tissue after mechanical mincing of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service