Skip to Content
Merck
All Photos(2)

Key Documents

C9891

Sigma-Aldrich

Collagenase from Clostridium histolyticum

Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Quality Level

form

essentially salt-free, lyophilized powder

specific activity

≥125 CDU/mg solid
0.5-5.0 FALGPA units/mg solid

mol wt

68-130 kDa

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

solubility

TESCA buffer (50 mM TES, 0.36 mM Calcium chloride, pH 7.4): soluble

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

The enzyme has also been used along with other proteases for the disaggregation of human tumor, mouse kidney, human brain, and lung epithelial tissues. It is also useful in liver and kidney perfusion studies, digestion of pancreas, isolation of nonparenchymal rat liver cells and hepatocytes. The enzyme from Sigma has been used for the digestion of type I collagen from bovine trabecular and cortical bones. It has also been used as a positive control in FALGPA assay on Group B Streptococci cells.

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum ranges from 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL are used for digestion. The duration of reaction varies from 15 minutes to several hours for a satisfactory efficiency of cell dissociation without causing too much cell death. Krebs Ringer buffer with calcium and BSA is preffered and Zn2+ is required for activity. The enzyme recognizes the sequence -R-Pro-8-X-Gly-Pro-R-, where X is most often a neutral amino acid. Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid (EGTA)4; β-mercaptoethanol; glutathione (reduced); thioglycolic acid, sodium; 2,2′-dipyridyl; 8-hydroxyquinoline, cysteine are known to inhibit the enzyme activity.

Caution

As supplied, this product is stable for one year at -20°C. There is no loss in FALGPA or protease activity in 30 days at 37°C, 50°C and -20°C. Solutions of crude collagenase are stable if frozen quickly in aliquots (at 10 mg/mL) and kept frozen at -20°C. Further freeze-thaw cycles will damage the solution. The product retains 100% activity over 7 hours when held on ice.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Preparation Note

This product is equivalent to first 40% ammonium sulfate fraction of Mandl, I., et al., J. Clin. Invest., 32, 1323 (1953). Solutions are typically prepared at 1-2 mg/mL in TESCA buffer (containing 50 mM TES, 0.36 mM Calcium chloride, pH 7.4 at 37°C.

substrate

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swati Sharma et al.
PloS one, 14(6), e0218194-e0218194 (2019-06-22)
From a biological and clinical perspective, it is imperative to establish primate spermatogonial cultures. Due to limited availability of human testicular tissues, the macaque (Macaca fascicularis) was employed as non-human primate model. The aim of this study was to characterize
C Fledelius et al.
The Journal of biological chemistry, 272(15), 9755-9763 (1997-04-11)
The heterogeneity of urinary degradation products of C-terminal telopeptides derived from the alpha1 chain of human type I collagen was investigated and characterized. The urinary fragments characterized in this study consisted of two cross-linked (X) amino acid sequences derived from
R J Jackson et al.
Infection and immunity, 62(12), 5647-5651 (1994-12-01)
Group B streptococci (GBS) are important pathogens in neonatal sepsis, pneumonia, and meningitis. The ability of GBS to invade the collagen-rich amniotic membrane of the placenta has been shown in vitro. In the presence of GBS, the collagen fibrils of
Alison E Casey et al.
The Journal of cell biology, 217(8), 2951-2974 (2018-06-21)
The mammary epithelium depends on specific lineages and their stem and progenitor function to accommodate hormone-triggered physiological demands in the adult female. Perturbations of these lineages underpin breast cancer risk, yet our understanding of normal mammary cell composition is incomplete.
Allison M L Nixon et al.
Scientific reports, 9(1), 842-842 (2019-01-31)
Cell surface antigen discovery is of great interest for biomedical research both for isolation of rare cell populations and therapeutic targeting. We developed a rapid, cost-effective, fully in vitro technology which facilities the simultaneous target discovery and human antibody generation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service