Skip to Content
Merck
All Photos(1)

Key Documents

21053

Sigma-Aldrich

Calcium phosphate monobasic monohydrate

purum p.a., ≥80% (KT)

Synonym(s):

Calcium bis(dihydrogenphosphate) monohydrate, Calcium dihydrogenphosphate monohydrate, Primary calcium phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(H2PO4)2 · H2O
CAS Number:
Molecular Weight:
252.07
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥80% (KT)
form:
powder or crystals

grade

purum p.a.

Quality Level

Assay

≥80% (KT)

form

powder or crystals

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: 100 mg/kg
K: ≤100 mg/kg
Na: 500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.[Ca++].OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Ca.2H3O4P.H2O/c;2*1-5(2,3)4;/h;2*(H3,1,2,3,4);1H2/q+2;;;/p-2

InChI key

ZBZJARSYCHAEND-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium phosphate monobasic monohydrate is mainly used in the manufacture of fertilizers and as an acidulant in food industry. Aqueous solution of calcium phosphate monobasic monohydrate (MCPM) mixed with α-tertiary calcium phosphate forms dicalcium phosphate dihydrate cement. It is also used in composing brushite cement.

Application

Calcium phosphate monobasic monohydrate may be used in the preparation of calcium polyphosphate (CPP) powders.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective formulations for the preparation of calcium phosphate bone cements.
Driessens FCM, et al.
Journal of Materials Science. Materials in Medicine, 5(3), 164-170 (1994)
Calcium phosphates in pharmaceutical tableting.
Schmidt PC and Herzog R.
Pharmacy World & Science : PWS, 15(3), 116-122 (1993)
Porous calcium polyphosphate scaffolds for bone substitute applications-in vitro characterization.
Pilliar RM, et al.
Biomaterials, 22(9), 963-972 (2001)
The Merck Index: An Encyclopedia of Chemicals, Drugs, and Biologicals
Royal Society of Chemistry null
Dan Meng et al.
Materials science & engineering. C, Materials for biological applications, 47, 266-272 (2014-12-11)
Calcium phosphate cements (CPCs) have been widely used as bone graft substitutes. However, the undesirable osteoinductivity and slow degradability of CPCs greatly hamper their clinical application. The aim of this study was to synthesize a type of injectable, bioactive cement.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service