Skip to Content
Merck
All Photos(3)

Documents

322431

Sigma-Aldrich

Potassium pyrophosphate

97%

Synonym(s):

Potassium diphosphate tetrabasic, Tetrapotassium diphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K4P2O7
CAS Number:
Molecular Weight:
330.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

1300 °C (lit.)

SMILES string

[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4K.H4O7P2/c;;;;1-8(2,3)7-9(4,5)6/h;;;;(H2,1,2,3)(H2,4,5,6)/q4*+1;/p-4

InChI key

RYCLIXPGLDDLTM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium pyrophosphate (K4P2O7) is a water soluble, buffering and chelating agent. It can also be used as an ionic dispersant to form a low viscous solution. It is a biocompatible and a non-toxic compound that is commonly used as a food additive.

Application

K4P2O7 can be used in the preparation of an electrolytic solution, which can further be used in the fabrication of lithium ion batteries and arrays of copper nanowires.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a base set of toxicity tests using ultrafine TiO2 particles as a component of nanoparticle risk management
Warheit DB, et al.
Toxicology Letters, 171(3), 99-110 (2007)
Stable high areal capacity lithium-ion battery anodes based on three-dimensional Ni-Sn nanowire networks
Tian M, et al.
Journal of Power Sources, 211(32), 46-51 (2012)
Electrochemical exfoliation of graphite to produce graphene using tetrasodium pyrophosphate
Kumar MKP, et al.
Royal Society of Chemistry Advances, 5(32), 24846-24852 (2015)
High clay content nanocomposite hydrogels with surprising mechanical strength and interesting deswelling kinetics
Liu Y, et al.
Polymer, 47(1), 1-5 (2006)
Copper nanowire arrays surface wettability control using atomic layer deposition of TiO 2
Abdulagatov AI, et al.
Russian Journal of Applied Chemistry, 89(8), 1265-1273 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service