Skip to Content
Merck
All Photos(1)

Key Documents

13204

Sigma-Aldrich

β-tri-Calcium phosphate

puriss. p.a., ≥98% β-phase basis (unsintered powder)

Synonym(s):

β-TCP, β-Tricalcium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca3O8P2
CAS Number:
Molecular Weight:
310.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥98% β-phase basis (unsintered powder)
form:
powder

grade

puriss. p.a.

Quality Level

Assay

≥98% β-phase basis (unsintered powder)

form

powder

impurities

≤250 mg/kg total sulfur (as SO4) (ICP)
≤50 mg/kg total heavy metals (as lead)

anion traces

chloride (Cl-): ≤100 mg/L

cation traces

As: ≤0.5 mg/kg
Ba: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤5 mg/kg
Cu: ≤1 mg/kg
Hg: ≤0.5 mg/kg
Ni: ≤1 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

SMILES string

[Ca+2].[Ca+2].[Ca+2].[P](=O)([O-])([O-])[O-].[P](=O)([O-])([O-])[O-]

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

InChI key

QORWJWZARLRLPR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

β-tri-Calcium phosphate belongs to the category of calcium-phosphate based ceramics, which has been involved in the preparation of a new biosynthetic material possessing morphogenetic activity that is used as a bone implant.

Analysis Note

Phase Composition (by XRD analysis)
Low crystalinity Hydroxylapatite at unsintered condition.
A high crystalinity ß-Tricalcium phosphate after sintering at 1100C.
ß-TCP: >98% (by x-ray diffraction analysis)
other Ca-P phases as Hydroxylapatite, a-TCP and Pyrophosphate <2.0%

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new polyphosphate calcium material with morphogenetic activity
Muller.G.EW, et al.
Materials Letters, 148, 163-166 (2015)
Timo Brandenburger et al.
Shock (Augusta, Ga.), 42(3), 234-238 (2014-07-01)
Remote ischemic preconditioning (RIPC) is an easily applicable method for protecting the heart against a subsequent ischemia and reperfusion (I/R) injury. However, the exact molecular mechanisms underlying RIPC are unknown. We examined the involvement of microRNAs (miRNAs) and in particular
Krzysztof W Luczynski et al.
Journal of biomedical materials research. Part A, 101(1), 138-144 (2012-07-25)
This paper is concerned with reliable and physically sound elasticity determination of rapid-prototyped tissue engineering scaffolds made of poly-L-lactide (PLLA), with and without small portions of tricalcium phosphate (TCP) inclusions. At the level of overall scaffolds, that is, that of
Floor C J van de Watering et al.
Journal of controlled release : official journal of the Controlled Release Society, 164(3), 283-290 (2012-07-18)
Clinical application of calcium phosphate cement (CPC; with incorporated polymeric porogens) in an injectable form implicates that loading methods for growth factors are limited. In view of this, the current study evaluated the in vitro and in vivo release kinetics
A H Choi et al.
Journal of dental research, 92(10), 853-859 (2013-07-17)
The purpose of coatings on implants is to achieve some or all of the improvements in biocompatibility, bioactivity, and increased protection from the release of harmful or unnecessary metal ions. During the last decade, there has been substantially increased interest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service