C7263
Calcium phosphate dibasic
98.0-105.0%
Synonym(s):
Calcium hydrogen phosphate
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
CaHPO4
CAS Number:
Molecular Weight:
136.06
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
98.0-105.0%
form
powder
SMILES string
[Ca++].OP([O-])([O-])=O
InChI
1S/Ca.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+2;/p-2
InChI key
FUFJGUQYACFECW-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Calcium phosphate dibasic has been used:
- to prepare high temperature stoichiometric sintered hydroxyapatite (sin-HA)
- as a component of calcium phosphate cement (CPC) to prepare bioartificial pancreas (BAP)
- to produce calcium pyrophosphate (Ca2P2O7) for the preparation of α-tricalcium phosphates (α-TCP)
Biochem/physiol Actions
Calcium phosphate dibasic is an anhydrate used to dilute tablets and capsules. It is widely used as an excipient for its compressibility, amenability, and brittle fracture propensity.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aditya M Kaushal et al.
Journal of pharmaceutical sciences, 100(4), 1456-1466 (2011-04-01)
Dibasic calcium phosphate occurs as an anhydrate (DCPA; CaHPO₄) and as a dihydrate (DCPD; CaHPO₄•2H₂O). Our objective was to investigate the unusual behavior of these phases. Dibasic calcium phosphate dihydrate was dehydrated in a (i) differential scanning calorimeter (DSC) in
Kai-Chiang Yang et al.
Transplantation, 90(6), 604-611 (2010-01-06)
Fibrous tissue outgrowth and hypoxia are the major restrictions for the application of bioartificial pancreas (BAP). Accordingly, the intramedullary cavity is proposed as an implant site, and a BAP constructed of calcium phosphate cement chamber was implanted. Mouse insulinoma cells
Anna Diez-Escudero et al.
Tissue engineering. Part C, Methods, 23(2), 118-124 (2017-01-14)
This article presents the application of dual focused ion beam/scanning electron microscopy (FIB-SEM) imaging for preclinical testing of calcium phosphates with osteoclast precursor cells and how this high-resolution imaging technique is able to reveal microstructural changes at a level of
C L Camiré et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 79(1), 159-165 (2006-04-15)
Alpha phase tricalcium phosphates (alpha-TCP) were produced using a solid-state reaction method and milled for various periods of time. The resulting four materials were alpha-TCPs, ranging in crystalline content. Powders were exposed to X-ray diffraction for material identification as well
Tae-Woo Kim et al.
Tissue engineering and regenerative medicine, 17(5), 607-624 (2020-08-18)
The delivery of growth factors using a carrier system presents a promising and innovative tool in tissue engineering and dentistry today. Two of the foremost bioactive factors, bone morphogenetic protein-2 and vascular endothelial growth factor (VEGF), are widely applied using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service