Skip to Content
Merck
All Photos(3)

Key Documents

307815

Sigma-Aldrich

Sodium metasilicate

Synonym(s):

Disodium metasilicate, Disodium silicate, Sodium silicate, Sodium silicon oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SiO3
CAS Number:
Molecular Weight:
122.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

composition

SiO2, 50-53%

Quality Level

SMILES string

[Na+].[Na+].[O-][Si]([O-])=O

InChI

1S/2Na.O3Si/c;;1-4(2)3/q2*+1;-2

InChI key

NTHWMYGWWRZVTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SMS is a silicon source for silica materials and can be used as a chemical intermediate for silica gel catalysts.

Application

Reactant for:
  • Hydrothermal preparation of thorium uranium silicate (Th1-xUxSiO4) uranothorite solid solutions
  • Fabrication of dense NaA zeolite coatings
  • Reactant for preparation of tungsten modified silica as catalyst for liquid phase oxidation of styrene
Sodium metasilicate (SMS) has been used in the preparation of polypyrrole/sodium metasilicate (PPy/Na2SiO3) and polythiophene/sodium metasilicate (PT/Na2SiO3) composites, novel high dielectric constant materials for use in power electronic systems.

SMS can also be used:
  • To prepare NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption and catalysis.
  • As a low-cost alternative to tetraethyl orthosilicate (TEOS) to synthesize highly ordered mesoporous silicas.
  • To prepare geopolymer concrete.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subramani Saravanabhavan et al.
Environmental science & technology, 39(10), 3776-3783 (2005-06-15)
A conventional dehairing process with sodium sulfide and lime is a major source of the pollution from the tanning industry. In other words, conventional dehairing processes degrade the hair to the extent that it cannot be recovered; thus, these processes
Highly porous silica networks derived from gelatin/siloxane hybrids prepared starting from sodium metasilicate
Retuert J, et al.
Journal of Non-Crystalline Solids, 347(1-3), 273-278 (2004)
Synthesis of highly ordered mesoporous silica materials using sodium silicate and amphiphilic block copolymers.
Kim J M and Stucky G D
Chemical Communications (Cambridge, England), 13, 1159-1160 (2000)
J Najda et al.
Biological trace element research, 42(1), 63-70 (1994-07-01)
The effect of an excessive inorganic silicon oral intake on the activity of basic antioxidant enzymes was studied in rats. Activities of superoxide dismutase, catalase, and glutathione peroxidase were measured in liver and kidney tissues of animals receiving per os
A Comparative Study on Strength Improvement of Concrete by Using Fly Ash Based Geopolymer.
Anand A, et al.
International Research Journal of Engineering and Technology (IRJET), 5(4), 4819-4821 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service