Skip to Content
Merck
All Photos(3)

Documents

C8017

Sigma-Aldrich

Calcium phosphate monobasic

≥95%

Synonym(s):

Calcium bis(dihydrogen phosphate), Calcium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(H2PO4)2
Molecular Weight:
234.05
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%

form

powder

solubility

H2O: slightly soluble(lit.)

SMILES string

[Ca++].OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Ca.2H3O4P/c;2*1-5(2,3)4/h;2*(H3,1,2,3,4)/q+2;;/p-2

InChI key

YYRMJZQKEFZXMX-UHFFFAOYSA-L

Related Categories

Application

Calcium phosphate monobasic can be used as starting material to prepare biomedical cements such as calcium phosphate silicate bone cement (CPSC) and calcium phosphate coatings on ceramic implants.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zubin Zhou et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(8), 2827-2840 (2017-12-29)
The influence of calcium polyphosphate (CPP) gel incorporation on the release of vancomycin and tobramycin from polymethyl methacrylate (PMMA) cement (Simplex P, SP) has been studied. Adding 10% CPP gel to SP led to a much lower burst release of
Irfan Qayoom et al.
Biomedical materials (Bristol, England), 15(5), 055015-055015 (2020-04-10)
Withania somnifera (ashwagandha) is used in Indian traditional medicine for its various health benefits. Withaferin-A, a steroidal lactone present in this herb, has shown proteosomal inhibition-based enhancement of bone mineralization. In the present work, chitosan microparticles blended with total methanolic
Pierre Schembri-Wismayer et al.
Journal of endodontics, 43(5), 751-759 (2017-03-16)
Tricalcium silicate (TCS)-based materials are used in endodontics because they are hydraulic and interact with blood, tissue fluids, and phosphate-based root canal irrigants, resulting in biomineralization. Newer-generation materials are biphasic; calcium phosphate is added to the TCS; thus, the material
Feiyue Li et al.
Chemosphere, 251, 126335-126335 (2020-03-08)
Pyrolysis of biomass with phosphate compound is a promising method to improve biochar characteristics. However, how phosphate compound affects the three components of biomass during the biochar formation is still unclear. In this study, a typical phosphate compound, calcium dihydrogen
Ivana Milanovic et al.
Journal of endodontics, 46(4), 515-523 (2020-02-26)
Open pores between endodontic sealer and root canal walls present potential niches for bacterial growth and migration pathways. The aim was to assess the internal and external porosity of calcium silicate-based sealers and to ascertain the long-term effect of storage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service