Skip to Content
Merck
All Photos(3)

Key Documents

E0396

Sigma-Aldrich

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid

BioXtra, ≥97 .0%

Synonym(s):

EGTA, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2OCH2CH2N(CH2CO2H)2]2
CAS Number:
Molecular Weight:
380.35
Beilstein:
1717370
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

product line

BioXtra

Quality Level

Assay

≥97 .0%

form

powder

reaction suitability

reagent type: chelator

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

241 °C (dec.) (lit.)

solubility

1 M NaOH: 0.1 M, clear, colorless
H2O: soluble (increases as pH is increased)

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

DEFVIWRASFVYLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A chelating agent useful for the determination of calcium in the presence of magnesium.

Biochem/physiol Actions

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid A is a chelating agent used for the determination of calcium in the presence of magnesium.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter Marcotti et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(16), 5505-5514 (2014-04-18)
Tip links between adjacent stereocilia are believed to gate mechano-electrical transducer (MET) channels and mediate the electrical responses of sensory hair cells. We found that mouse auditory hair cells that lack tip links due to genetic mutations or exposure to
David R Whitfield et al.
The American journal of geriatric psychiatry : official journal of the American Association for Geriatric Psychiatry, 23(2), 141-148 (2014-06-24)
Depression is a common symptom in dementia with Lewy bodies (DLB), Parkinson disease dementia (PDD), and Alzheimer disease (AD), yet its molecular basis remains unclear and current antidepressants do not appear to be effective. Cerebral zinc has been implicated in
Olof Idevall-Hagren et al.
The EMBO journal, 34(17), 2291-2305 (2015-07-24)
The extended synaptotagmins (E-Syts) are ER proteins that act as Ca(2+)-regulated tethers between the ER and the plasma membrane (PM) and have a putative role in lipid transport between the two membranes. Ca(2+) regulation of their tethering function, as well
Jennifer R Brum et al.
Science (New York, N.Y.), 348(6237), 1261498-1261498 (2015-05-23)
Viruses influence ecosystems by modulating microbial population size, diversity, metabolic outputs, and gene flow. Here, we use quantitative double-stranded DNA (dsDNA) viral-fraction metagenomes (viromes) and whole viral community morphological data sets from 43 Tara Oceans expedition samples to assess viral
Anita C Bellail et al.
Nature communications, 5, 4234-4234 (2014-06-24)
Ubiquitination governs oscillation of cyclin-dependent kinase (CDK) activity through a periodic degradation of cyclins for orderly cell cycle progression; however, the mechanism that maintains the constant CDK protein levels throughout the cell cycle remains unclear. Here we show that CDK6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service