Skip to Content
Merck
All Photos(6)

Key Documents

A2383

Sigma-Aldrich

Adenosine 5′-triphosphate disodium salt hydrate

Grade I, ≥99%, from microbial

Synonym(s):

ATP disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5Na2O13P3 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

microbial

Quality Level

type

Grade I

Assay

≥99%

form

powder

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate disodium salt hydrate has been used:
  • to prepare adenosine triphosphate (ATP) standard solutions to determine ATP levels in various bacterial cultures
  • bone marrow-derived macrophages (BMDM) treatment
  • as a preparation free of contaminating guanine nucleotides to study the binding effects of coatomer β subunit (β-COP)

Biochem/physiol Actions

Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, active transport, and motility. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase that produce the secondary messenger cyclic adenosine monophosphate (cAMP). ATP provides the metabolic energy to drive metabolic pumps. It serves as a coenzyme in a wide array of enzymatic reactions.
P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems
P2 purinocepter agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physiology, Adenosine Triphosphate (ATP)
NCBI Bookshelf null
Mass spectrometry and NMR spectroscopy based quantitative metabolomics
Proteomics. Clinical Applications (2020)
Release of extracellular ATP by bacteria during growth
Mempin R, et al.
BMC Microbiology, 13(1), 301-301 (2013)
J G Donaldson et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(14), 6408-6412 (1992-07-15)
The coatomer is a cytosolic protein complex that reversibly associates with Golgi membranes and is implicated in modulating Golgi membrane transport. The association of beta-COP, a component of coatomer, with Golgi membranes is enhanced by guanosine 5'-[gamma-thio]triphosphate (GTP[gamma S]), a
Rui Nian et al.
Biotechnology progress, 25(4), 1078-1085 (2009-06-25)
The use of polyethylene glycol (PEG) as a refolding additive to a refolding cocktail comprising the molecular bichaperone ClpB and DnaKJE significantly enhances chaperone-mediated refolding of heat-denatured malate dehydrogenase (MDH). The critical factor to affect the refolding yield is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service