Skip to Content
Merck
All Photos(4)

Key Documents

A4926

Sigma-Aldrich

1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid

98%

Synonym(s):

2,2′-(Ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid, BAPTA, Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O10
CAS Number:
Molecular Weight:
476.43
Beilstein:
5192406
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

storage temp.

2-8°C

SMILES string

OC(=O)CN(CC(O)=O)c1ccccc1OCCOc2ccccc2N(CC(O)=O)CC(O)=O

InChI

1S/C22H24N2O10/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32)

InChI key

FTEDXVNDVHYDQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In nature 1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetra acetic acid (BAPTA) is a main metabolic product of BAPTA -AM (BAPTA-tetra(actoxymethyl ester), a neuro protective agent in cerebral ischemia, in rats.BAPTA is an intercellular Ca2+ chelator.

Application

Used to study morphological and signaling events of Ca2+ deficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siu-Kei Chow et al.
ASN neuro, 2(1), e00026-e00026 (2009-12-17)
The contribution of astrocytes to the pathophysiology of AD (Alzheimer's disease) and the molecular and signalling mechanisms that potentially underlie them are still very poorly understood. However, there is mounting evidence that calcium dysregulation in astrocytes may be playing a
Zheng Feng et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(30), 3052-3058 (2010-10-23)
BAPTA free acid was identified as the main metabolic product of 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetra(actoxymethyl ester) (BAPTA-AM), a neuroprotective agent in cerebral ischemia, in rats. In this paper, liquid chromatography-ultraviolet (LC-UV) and mass spectrometry/mass spectrometry (LC-MS/MS) methods were employed for the
K S Han et al.
Journal of neurochemistry, 78(2), 230-239 (2001-07-20)
Sustained alteration in [Ca(2+)]i triggers neuronal death. We examined morphological and signaling events of Ca(2+)-deficiency-induced neuronal death. Cortical cell cultures exposed to 20 microM 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid (BAPTA-AM), an intracellular calcium chelator, underwent neuronal apoptosis within 12 h that was evident
Mayeul Collot et al.
Journal of the American Chemical Society, 134(36), 14923-14931 (2012-07-24)
We designed Calcium Rubies, a family of functionalizable BAPTA-based red-fluorescent calcium (Ca(2+)) indicators as new tools for biological Ca(2+) imaging. The specificity of this Ca(2+)-indicator family is its side arm, attached on the ethylene glycol bridge that allows coupling the
Roger C Hardie
Cell calcium, 38(6), 547-556 (2005-09-06)
In vivo light-induced and basal hydrolysis of phosphatidyl inositol 4,5-bisphosphate (PIP2) by phospholipase C (PLC) were monitored in Drosophila photoreceptors using genetically targeted PIP2-sensitive ion channels (Kir2.1) as electrophysiological biosensors for PIP2. In cells loaded via patch pipettes with varying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service