Skip to Content
Merck
All Photos(6)

Key Documents

RDD002

Sigma-Aldrich

HEPES

≥99.5%, crystalline powder, anhydrous, Redi-Dri

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Product Name

HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.5%

form

crystalline powder

quality

free-flowing

pH

5.0-6.5 (25 °C, 238 g/L)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 500 mg/mL, clear, colorless

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.

Application

HEPES has been used:
  • In the novel vesicle-budding protocol as a component in stock solution and buffer solutions
  • As a component in intracellular solution for the preparation of brain-slice recording
  • As a component of wash buffer, elution buffer and desalting buffer during affinity protein purification
  • To maintain the pH at 7 in thermal denaturation assay of purified protein
  • In isocratic elution of protein using gel filtration
  • Along with NaCl in sample preparation for EPR (Electron Paramagnetic Resonance) spectroscopy

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell-free Generation of COPII-coated Procollagen I Carriers.
Yuan L, et al.
Bio-protocol, 7(22) (2017)
Discovery and characterization of a prevalent human gut bacterial enzyme sufficient for the inactivation of a family of plant toxins.
Koppel N, et al.
eLife, 7, e33953-e33953 (2018)
Optogenetic Mapping of Synaptic Connections in Mouse Brain Slices to Define the Functional Connectome of Identified Neuronal Populations.
Mingote S, et al.
The Journal of Neuroscience (2015)
Assessing the corrosion of biodegradable magnesium implants: a critical review of current methodologies and their limitations.
Kirkland NT, et al.
Acta Biomaterialia, 8(3), 925-936 (2012)
Hydrogen ion buffers for biological research.
Good NE, et al.
Biochemistry, 5(2), 467-477 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service