Skip to Content
Merck
All Photos(6)

Key Documents

G8877

Sigma-Aldrich

Guanosine 5′-triphosphate sodium salt hydrate

Guanosine 5′-triphosphate sodium salt hydrate

≥95% (HPLC), powder

Synonym(s):

GTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+
CAS Number:
Molecular Weight:
523.18 (free acid basis)
Beilstein:
3585677
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H16N5O14P3.3Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1

InChI key

KZRMTEVIDYXWQW-CYCLDIHTSA-K

Gene Information

human ... HRAS(3265)

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine 5′-triphosphate sodium salt hydrate has been used:
  • in the preparation of horseradish peroxidase (HRP)-modified platination reagents[1]
  • in whole cell, voltage-clamp recordings[2]
  • in in vitro inhibition of transglutaminase (tTG)[3]

Biochem/physiol Actions

GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

Prepared by enzymatic phosphorylation of 5′-GMP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T cells from celiac disease lesions recognize gliadin epitopes deamidated in situ by endogenous tissue transglutaminase
Molberg O, et al.
European Journal of Immunology, 31(5), 1317-1323 (2001)
Feng Rao et al.
Analytical biochemistry, 389(2), 138-142 (2009-03-31)
The cyclic dinucleotide c-di-GMP is a widespread bacterial messenger molecule with potential application as a therapeutic agent for treating bacterial infection. Current enzymatic synthesis of c-di-GMP using mesophilic diguanylate cyclase (DGC) proteins suffers from low production yield due to protein
Flufenamic acid affects multiple currents and causes intracellular Ca2+ release in Aplysia bag cell neurons
Gardam K E, et al.
Journal of Neurophysiology, 100(1), 38-49 (2008)
Rob P M van Gijlswijk et al.
Clinical chemistry, 48(8), 1352-1359 (2002-07-27)
Microarray platforms will change immunochemical and nucleic acid-based analysis of cell homogenates and body fluids compared with classic analyses. Microarrays use labeled target and immobilized probes, rather than fixed targets and labeled probes. We describe a method for simultaneous labeling
M Uriarte et al.
The Biochemical journal, 293 ( Pt 1), 93-100 (1993-07-01)
A glycoprotein fraction was isolated from rat liver membranes by affinity chromatography on immobilized wheat-germ lectin. Incubation of this fraction with MgATP or MgGTP resulted in a sequential phosphorylation and dephosphorylation of a complex of three polypeptides (118, 128 and

Questions

  1. Can assistance be provided in sourcing ATP and GTP for establishing an in vitro translation system from plant cells?

    1 answer
    1. Upon verification, there is currently no validated ATP or GTP product for in vitro translation application available. However, according to the following publication, ATP (cat#A2383) and GTP (cat#G8877) seem to work well for in vitro transcription and translation systems.

      Reference: Polyvinylsulfonic acid: A Low-cost RNase inhibitor for enhanced RNA preservation and cell-free protein translation

      Bioengineered. 2018 Jan 1;9(1):90-97. doi: 10.1080/21655979.2017.1313648. Epub 2017 Jun 29.PMID: 28662363

      Helpful?

Reviews

Active Filters

    • Reviews 2
    • Votes 0
    5 out of 5 stars.

    It is a high quality product and can be use for research purposes.

    Excellent product and I use it to measure GTPase activity.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service