Skip to Content
Merck
All Photos(1)

Key Documents

SLM-241

Sigma-Aldrich

DMEM - high glucose

Complete medium, with ʟ-glutamine and sodium pyruvate and 10% FBS, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, DMEM with 10% FBS, Dulbecco′s Modified Eagle′s Medium - high glucose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Product Name

DMEM Complete Medium, with 2mM L-Glut, Sodium Pyruvate, and 10% FBS, contains DMEM high glucose medium with 2mM L-glut, sodium pyruvate, and 10% U.S. origin FBS

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

General description

Dulbecco′s Modified Eagle (DMEM) Complete Medium contains DMEM high glucose medium with 2mM L-glut, sodium pyruvate, and 10% U.S. origin FBS. DMEM Complete Medium is a convenient sterile solution for the culture of primary mammalian cells including mouse and chicken cells, and a variety of normal and transformed cells.

Application

DMEM Complete Medium, with 2mM L-Glut, Sodium Pyruvate, and 10% FBS has been used:
  • to culture human gingival fibroblasts
  • to culture mouse fibroblast cells
  • as an overlay medium for severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) plaque assay

Quality

pH= 7.2-7.5
Osmolality= 320-346
Endotoxin= ≤ 2EU/mL
Mycoplasma= Negative
Sterility Tested= Negatvie for Growth
Appearance= Orange/ Red Clear Liquid
Cell Line Tested= HT-22 Human Neuronal Cells

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytotoxicity and Collagen Expression Effects of Tideglusib Administration on Human Periodontal Cells: An In-Vitro Study
Buse ONCU, et al.
Clinical and Experimental Medical Sciences, 10(2), 153-162 (2020)
Leonard F Blackwell et al.
Frontiers in public health, 6, 153-153 (2018-06-16)
The patterns of a woman's normal ovarian activity can take many forms from childhood to menopause. These patterns lie on a continuum ranging from no ovarian activity to a fully fertile ovulatory cycle, but among the other defined patterns are
Sumaira Javaid et al.
PloS one, 14(11), e0225056-e0225056 (2019-11-20)
Natural flora is the richest source of novel therapeutic agents due to their immense chemical diversity and novel biological properties. In this regard, eighteen natural products belonging to different chemical classes were evaluated for their thymidine phosphorylase (TP) inhibitory activity.
Juan A Perez-Bermejo et al.
Science translational medicine, 13(590) (2021-03-17)
Although coronavirus disease 2019 (COVID-19) causes cardiac dysfunction in up to 25% of patients, its pathogenesis remains unclear. Exposure of human induced pluripotent stem cell (iPSC)-derived heart cells to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) revealed productive infection and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service