Skip to Content
Merck

Skip To

D6546

DMEM - high glucose

With sodium Bicarbonate, sodium pyruvate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Dulbecco’s Modified Eagle’s Medium - high glucose
1 of 2 reviewers received a sample product or took part in a promotion

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
phenol red: yes
sodium pyruvate: yes
HEPES: no
L-glutamine: no
glucose: high

shipped in

ambient

storage temp.

2-8°C

General description

This DMEM-Hi glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Preparation Note

Supplement with 0.584 g/L L-glutamine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D6429D5796D5671
sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

components

NaHCO3: yes
phenol red: yes
sodium pyruvate: yes
HEPES: no
L-glutamine: no
glucose: high

components

HEPES: no
NaHCO3: yes
phenol red: yes
sodium pyruvate: yes
L-glutamine: yes
glucose: high

components

NaHCO3: yes
L-glutamine: yes
glucose: high
phenol red: yes
sodium pyruvate: no
HEPES: no

components

L-glutamine: no
glucose: high
NaHCO3: yes
sodium pyruvate: no
phenol red: yes
HEPES: no

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Iman Azimi et al.
Scientific reports, 7(1), 15140-15140 (2017-11-11)
One of the hallmarks of the tumour microenvironment is hypoxia resulting from increased oxygen consumption by proliferative cancer cells and altered vasculature. Hypoxic tension initiates various cellular signals and can drive epithelial to mesenchymal transition (EMT), a process important in
Gloria Bonuccelli et al.
Oncotarget, 8(13), 20667-20678 (2017-02-23)
Here, we assembled a broad molecular "tool-kit" to interrogate the role of metabolic heterogeneity in the propagation of cancer stem-like cells (CSCs). First, we subjected MCF7 cells to "metabolic fractionation" by flow cytometry, using fluorescent mitochondrial probes to detect PCG1α
Morag R Hunter et al.
Journal of molecular biology, 430(14), 2153-2163 (2018-05-21)
Multi-subunit tethering complexes control membrane fusion events in eukaryotic cells. Class C core vacuole/endosome tethering (CORVET) and homotypic fusion and vacuole protein sorting (HOPS) are two such complexes, both containing the Sec1/Munc18 protein subunit VPS33A. Metazoans additionally possess VPS33B, which



Global Trade Item Number

SKUGTIN
D6546-24X500ML04061833587768
D6546-6X500ML04061833587782
D6546-500ML04061833587775

Questions

  1. Which DMEM media and glutamine products would you recommend for the PEA-13-97040311 cell line purchased from ECACC?

    1 answer
    1. According to ECACC, D6546 (DMEM) and G7513 (glutamine) are on the recommended product list for reagents required in cell culture applications.

      Helpful?

Reviews

Active Filters

  1. London, UK
    • Review 1
    • Votes 3
    5 out of 5 stars.

    Things back to normal

    After the shortages because of Brexit/Covid its great everything is getting back to normal

    Helpful?

  2. 1 Ratings-only review