Skip to Content
Merck
All Photos(1)

Key Documents

171260

Sigma-Aldrich

Dorsomorphin

≥95% (HPLC), solid, AMPK inhibitor, Calbiochem

Synonym(s):

AMPK Inhibitor, Compound C, 6-[4-(2-Piperidin-1-yl-ethoxy)-phenyl)]-3-pyridin-4-yl-pyrrazolo[1,5-a]-pyrimidine, Dorsomorphin, BMP Inhibitor I, AMPK Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25N5O
CAS Number:
Molecular Weight:
399.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

AMPK Inhibitor, Compound C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC₅₀ = 25.1, 148, and 234.6 nM, respectively).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

2% HCl: 10 mg/mL
2% acetic acid: 10 mg/mL
methanol: 3 mg/mL
DMSO: 4 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21ncc(c2ncc(c1)c4ccc(cc4)OCCN5CCCCC5)c3ccncc3

InChI

1S/C24H25N5O/c1-2-12-28(13-3-1)14-15-30-22-6-4-19(5-7-22)21-16-26-24-23(17-27-29(24)18-21)20-8-10-25-11-9-20/h4-11,16-18H,1-3,12-15H2

InChI key

XHBVYDAKJHETMP-UHFFFAOYSA-N

General description

A cell-permeable pyrrazolopyrimidine compound that inhibits against KDR/VEGFR2, ALK2/BMPR-I, AMPK kinase activity (IC50 = 25.1, 148, and 234.6 nM, respectively), while exhibiting much reduced or little effect toward ALK5/TGFβR-I, ZAPK, Syk, PKCθ, PKA, or JAK3. Shown to block both BMP pathway-dependent dorsoventral development (EC100 = 2.5 µM) and VEGF signaling-dependent intersomitic vessel formation (EC50 = 5 µM) in zebrafish embryo in vivo. Commonly used in combination with AMPK activators AICAR (Cat. No. 123040) and/or Metformin (Cat. No. 317240) for studying AMPK-dependent cellular events in vitro and physiological responses in animals in vivo. Also available as a 10 mM solution in DMSO (Cat. No. 171261).

Biochem/physiol Actions

Cell permeable: yes
EC100 = 2.5 µM against BMP pathway-dependent dorsoventral development; EC50 = 5 µM against VEGF signaling-dependent intersomitic vessel formation in zebrafish embryo in vivo
Primary Target
AMPK
Product competes with ATP.
Reversible: yes
Target IC50: 25.1, 148, and 234.6 nM, against KDR/VEGFR2, ALK2/BMPR-I, AMPK kinase activity, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hao, J., et al. 2010. ACS Chem.Biol.5, 245.
Yu, P.B., et al. 2008. Nat. Chem. Biol4, 33.
Kim, E.K., et al. 2004. J. Biol. Chem.279, 19970.
Lee, M., et al. 2003. J. Biol. Chem.278, 39653.
Inoki, K., et al. 2003. Cell115, 577.
Fryer, L.G., 2002. FEBS Lett.531, 189.
Zhou, G., et al. 2001. J. Clin. Invest.108, 1167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamadreza Savaee et al.
Reports of biochemistry & molecular biology, 11(1), 157-165 (2022-06-30)
Prostate cancer is known as one of the most prevalent health disorders in the male population globally. The aim of the current study was to evaluate the effects of separate and concomitant use of MK-2206 and salinomycin on prostate cancer
Jian Wen et al.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology, 32(1), 97-105 (2021-04-25)
Bile acid metabolism is a contributing factor that promotes cholelithiasis. Recent studies have suggested novel roles of leptin in the formation of gallbladder stones (GS); however, no evidence confirmed the function of leptin in the formation of primary intrahepatic bile
Tae Suk Kim et al.
International journal of molecular sciences, 22(18) (2021-09-29)
The Warburg effect is important for cancer cell proliferation. This phenomenon can be flexible by interaction between glycolysis and mitochondrial oxidation for energy production. We aimed to investigate the anticancer effects of the pyruvate dehydrogenase kinase inhibitor, dichloroacetate (DCA) and
Weixin Li et al.
Immunopharmacology and immunotoxicology, 43(4), 431-442 (2021-06-24)
Acute lung injury (ALI) is a devastating lung disease characterized by uncontrolled pulmonary inflammation and oxidative stress. Currently, no effective therapeutic strategies are available for ALI and its prognosis remains poor. The present study aims to investigate the role and
Dong Liang et al.
Scientific reports, 6, 21914-21914 (2016-02-24)
Mesenchymal stem cells (MSCs)-based therapy provides a promising avenue for the management of peripheral arterial disease (PAD). However, engrafted MSCs are subjected to acute cell death in the ischemic microenvironment. Apelin has been shown to protect bone marrow MSCs against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service