Skip to Content
Merck
All Photos(2)

Documents

A1393

Sigma-Aldrich

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate

≥93%

Synonym(s):

AICAR monophosphate, N1-(β-D-5′-Phosphoribofuranosyl)-5-aminoimidazole-4-carboxamide, NSC 283955, NSC 292227, ZMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N4O8P
CAS Number:
Molecular Weight:
338.21
EC Number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥93%

form

powder

mol wt

338.21  g/mol

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O[C@@H]1COP(O)(O)=O

InChI

1S/C9H15N4O8P/c10-7-4(8(11)16)12-2-13(7)9-6(15)5(14)3(21-9)1-20-22(17,18)19/h2-3,5-6,9,14-15H,1,10H2,(H2,11,16)(H2,17,18,19)/t3-,5-,6-,9-/m1/s1

InChI key

NOTGFIUVDGNKRI-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranosyl 5′-monophosphate (AICAR), a natural metabolic intermediate of purine biosynthesis, is usually found in all organisms. It is produced from succinyl-AICAR (SAICAR) with the help of adenylosuccinate lyase (ASL) enzyme.

Application

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate has been used in the study to explore the anticancer effect of nine statins in breast cancer and glioblastoma.
AICAR monophosphate is a 5′′-phosphorylated analog of cell permeable AICAR that mimics AMP. AICAR is an adenosine monophophate (AMP)-activated protein kinase (AMPK) activator/agonist.

Biochem/physiol Actions

A 5′-phosphorylated analog of cell permeable AICAR that mimics adenosine monophosphate (AMP). It is an AMP-activated protein kinase (AMPK) activator.
In humans, 5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate (AICAR) is found to be accumulated in numerous metabolic diseases. It can increase the endurance of sedentary mice. AICAR exhibits antiproliferative effects. It can induce apoptosis of aneuploid cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans P M M Lauritzen et al.
Diabetes, 62(9), 3081-3092 (2013-06-14)
Recent studies suggest that interleukin 6 (IL-6) is released from contracting skeletal muscles; however, the cellular origin, secretion kinetics, and signaling mechanisms regulating IL-6 secretion are unknown. To address these questions, we developed imaging methodology to study IL-6 in fixed
Hiroyasu Hatakeyama et al.
Molecular biology of the cell, 24(6), 809-817 (2013-01-18)
Tbc1d1 is key to skeletal muscle GLUT4 regulation. By using GLUT4 nanometry combined with a cell-based reconstitution model, we uncover a shift in the regulatory mode of Tbc1d1 by showing that Tbc1d1 temporally acquires insulin responsiveness, which triggers GLUT4 trafficking
Takashi Sasaki et al.
American journal of physiology. Endocrinology and metabolism, 306(9), E1085-E1092 (2014-03-20)
Exercise can effectively ameliorate type 2 diabetes and insulin resistance. Here we show that the mRNA levels of one of peroxisome proliferator-activated receptor (PPAR) family members, PPARγ1, and genes related to energy metabolism, including PPARγ coactivator-1 protein-1α (PGC-1α) and lipoprotein
Joshua D Stone et al.
American journal of physiology. Heart and circulatory physiology, 304(3), H369-H381 (2012-12-04)
Vascular smooth muscle cell (VSMC) activation promotes a synthetic phenotype that underlies many vessel growth disorders. In this regard it has been suggested that the metabolic sensor adenosine 5'-monophosphate-activated protein kinase (AMPK) has significant antigrowth and antimetastatic properties and may
Lykke Sylow et al.
Diabetes, 62(4), 1139-1151 (2013-01-01)
In skeletal muscle, the actin cytoskeleton-regulating GTPase, Rac1, is necessary for insulin-dependent GLUT4 translocation. Muscle contraction increases glucose transport and represents an alternative signaling pathway to insulin. Whether Rac1 is activated by muscle contraction and regulates contraction-induced glucose uptake is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service