R5010
Resveratrol
≥99% (HPLC)
Synonym(s):
3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
About This Item
Recommended Products
biological source
plant root (bushy knotweed)
Assay
≥99% (HPLC)
form
powder
solubility
acetone: 50 mg/mL, clear, faintly yellow
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
SMILES string
Oc1ccc(cc1)\C=C\c2cc(O)cc(O)c2
InChI
1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+
InChI key
LUKBXSAWLPMMSZ-OWOJBTEDSA-N
Gene Information
human ... AR(367) , CYP1A2(1544) , ESR1(2099) , SIRT1(23411)
mouse ... Nos2(18126)
rat ... Esr1(24890)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- in entrapment experiment
- to study its neuroprotective effect on retinal neuronal cell death mediated by diabetes-induced activation of Ca2+/calmodulin-dependent protein kinase II
- as a standard in the assessment of stilbene content in grapevine leaf by in vivo fluorometry
Biochem/physiol Actions
Preparation Note
also commonly purchased with this product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>392.0 °F - closed cup - (External MSDS)
Flash Point(C)
> 200.0 °C - closed cup - (External MSDS)
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service