Skip to Content
Merck
All Photos(1)

Key Documents

A5762

Sigma-Aldrich

Adenine 9-β-D-arabinofuranoside

≥99%

Synonym(s):

9-β-D-Arabinofuranosyladenine, Ara-A, Vidarabine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
Beilstein:
624881
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99%

form

powder

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7+,10-/m1/s1

InChI key

OIRDTQYFTABQOQ-UHTZMRCNSA-N

Gene Information

mouse ... Ahcy(269378)

Looking for similar products? Visit Product Comparison Guide

General description

Adenine 9-β-D-arabinofuranoside (AraA) is a nucleoside analog.[1] It is an antiviral drug and targets viral DNA polymerases[1] and is majorly used for treating herpes simplex viral infection.[2] It is a potent inhibitor of AMP-activated protein kinase (AMPK).[3] It also inhibits cardiac type 5 adenylyl cyclase but does not improve pathology in cardiovascular diseases.[2]

Application

Adenine 9-β-D-arabinofuranoside has been used for the inhibition of 5′ AMP-activated protein kinase (AMPK) in liver, [4] muscle[5] and cardiac cells H9c2.[6]
Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling.

Biochem/physiol Actions

Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 μM. Clinically significant antiviral agent, especially against herpes simplex (HSV), by inhibition of DNA polymerase.
Cell-permeable adenylyl cyclase inhibitor. IC50 = 30 μM in detergent-dispersed rat brain preparation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The direct effect of leptin on skeletal muscle thermogenesis is mediated by substrate cycling between de novo lipogenesis and lipid oxidation
Solinas G, et al.
Febs Letters, 577(3), 539-544 (2004)
GYY4137, a novel hydrogen sulfide-releasing molecule, likely protects against high glucose-induced cytotoxicity by activation of the AMPK/mTOR signal pathway in H9c2 cells
Wei W, et al.
Molecular and Cellular Biochemistry, 389(1-2), 249-256 (2014)
AMPK regulation of mouse oocyte meiotic resumption in vitro
Chen J, et al.
Developmental Biology, 291(2), 227-238 (2006)
AMPK involvement in endoplasmic reticulum stress and autophagy modulation after fatty liver graft preservation: a role for melatonin and trimetazidine cocktail
Zaouali MA, et al.
Journal of Pineal Research, 55(1), 65-78 (2013)
Antiviral drug vidarabine possessing cardiac type 5 adenylyl cyclase inhibitory property did not affect cardiohemodynamic or electrophysiological variables in the halothane-anesthetized dogs
Wada T, et al.
The Journal of Toxicological Sciences, 41(1), 115-122 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service