Skip to Content
Merck
All Photos(3)

Key Documents

PHOSS-RO

Roche

PhosSTOP

sufficient for 10 10mL buffer preparations, sufficient for 20 10mL buffer preparations, suitable for tissue processing, suitable for immunoprecipitation (IP)

Synonym(s):

inhibitor tablets for phosphatase, phosphatase inhibitor tablets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.28
form:
tablet
solubility:
aqueous buffer: soluble
storage temp.:
2-8°C

form

tablet

usage

sufficient for 10 10mL buffer preparations
sufficient for 20 10mL buffer preparations

packaging

pkg of 10 tablets (04906845001)
pkg of 20 tablets (04906837001)

manufacturer/tradename

Roche

technique(s)

immunoprecipitation (IP): suitable
tissue processing: suitable

solubility

aqueous buffer: soluble

storage temp.

2-8°C

General description

PhosSTOP tablets are a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use tablet which dissolves quickly in aqueous solutions (buffers), including buffers containing formalin (for formalin fixation).

Specificity

Protects proteins against dephosphorylation. Inhibits phosphatase classes such as acid and alkaline phosphatases, as well as serine/threonine (PP1, PP2A, and PP2B) and tyrosine protein phosphatases (PTP) in bacterial, mammalian, yeast, and plant extracts.

Application

PhosSTOP has been used as a component of
  • lysis buffer for the lysis of frozen mouse tissue to obtain protein supernatant
  • lysis of bone derived macrophages
  • nuclear extraction buffer for cell lysis and immunoprecipitation

PhosSTOP protects protein extracts against dephosphorylation. The tablets inhibit a broad spectrum of phosphatases such as acidic and alkaline phosphatases, as well as serine/threonine (PP1, PP2A, and PP2B), dual-specificity phosphatases and tyrosine protein phosphatases (PTP) in bacterial, insect, mammalian, yeast, and plant extracts. PhosSTOP Tablets are well suited for the purification and detection of phoshorylated proteins. They are also used to prevent dephosphorylation in formalin-fixed paraffin embedded (FFPE) tissue sections.
PhosSTOP tablets can be applied in combination with cOmplete Protease Inhibitor Cocktail Tablets to simultaneously protect proteins against dephosphorylation and proteolytic degradation. Both inhibitor tablets maintain their activity when used together.
PhosSTOP inhibitors were evaluated for their effect on protein assays. The inhibition shows almost no influence on BCA or Bradford protein assays.

Features and Benefits

Achieve immediate, effective, and convenient inhibition of a broad spectrum of phosphatases across a wide range of sample materials (e.g., mammalian, plant, yeast, and bacteria) with non-toxic PhosSTOP Phosphatase Inhibitor Cocktail Tablets. PhosSTOP is a proprietary blend of phosphatase inhibitors, formulated as a ready-to-use, quick-dissolving, water-soluble tablet.

  • Comprehensive protection: Inhibit acid and alkaline phosphatases, as well as serine/threonine (PP1, PP2A, and PP2B) and tyrosine protein phosphatases.
  • Safe: Dissolve the non-toxic PhosSTOP Tablets in aqueous solutions and avoid organic solvents (e.g., DMSO).
  • Suitable for formalin fixation: Maintain phosphorylation status of proteins in histological sections.
  • Convenient: Eliminate the time-consuming search for just the right phosphatase inhibitor.
  • Easy to use: Simply drop a quick-dissolving tablet into your buffer.
  • Stable: Store stock solutions for 1 month at +2 to +8°C, or for at least 6 months at -15 to -25°C.

For all-inclusive protein protection, combine PhosSTOP with cOmplete or cOmplete ULTRA Protease Inhibitor Tablets to inhibit both phosphatases and proteases.

Preparation Note

Working concentration: 1 tablet per 10 ml cell extract or buffer
Working solution: Solvent is recommended in distilled water or aqueous buffers (including buffers containing formalin).
Dissolve one tablet per 10 ml aqueous buffer. Alternatively, prepare a 10x stock solution in 1 ml dist. water or 100 mM phosphate buffer, pH 7.0. Higher concentrations of stock solutions (e.g., 20x) are possible.
Storage conditions (working solution): 2 to 8 °C

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

PhosSTOP is a trademark of Roche

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The SMX DNA repair tri-nuclease
Wyatt HDM, et al.
Molecular Cell, 65(5), 848-860 (2017)
Piliang Hao et al.
PloS one, 6(2), e16884-e16884 (2011-03-05)
Protein post-translational modifications (PTMs) are regulated separately from protein expression levels. Thus, simultaneous characterization of the proteome and its PTMs is pivotal to an understanding of protein regulation, function and activity. However, concurrent analysis of the proteome and its PTMs
Expression of siglec-E alters the proteome of lipopolysaccharide (LPS)-activated macrophages but does not affect LPS-driven cytokine production or toll-like receptor 4 endocytosis
Nagala M, et al.
Frontiers in Immunology, 8, 1926-1926 (2018)
Huperzine A ameliorates obesity-related cognitive performance impairments involving neuronal insulin signaling pathway in mice
Wang HY, et al.
Acta Pharmacologica Sinica, 1-1 (2019)
Fabio P S Santos et al.
Blood, 115(6), 1131-1136 (2009-12-17)
Few treatment options exist for patients with myelofibrosis (MF), and their survival is significantly shortened. Activating mutation of the JAK2 tyrosine kinase (JAK2(V617F)) is found in approximately 50% of MF patients. CEP-701 is a tyrosine kinase inhibitor that inhibits JAK2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service