Skip to Content
Merck
All Photos(1)

Key Documents

T-033

Supelco

exo-THC solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

exo-THC solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

−20°C

SMILES string

CCCCCc1cc(O)c2C3CC(=C)CC[C@H]3C(C)(C)Oc2c1

InChI

1S/C21H30O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h12-13,16-17,22H,2,5-11H2,1,3-4H3/t16?,17-/m1/s1

InChI key

AOYYFUGUUIRBML-ZYMOGRSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

exo-THC, also known as ∆9-11-THC, occurs as an impurity of ∆9-THC formed during its synthesis.

Application

The Certified Spiking Solution® is suitable for exo-THC testing by HPLC, GC/MS, or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and cannabis potency or impurity profiling testing applications. The Certified Reference Material (CRM) in solution can also be used as follows:
  • Analysis of 29 hemp-based veterinary products to measure the amount of cannabinoids, terpenes, and heavy metals by liquid chromatography-diode array detection/mass spectroscopy (LC-DAD/MS), headspace gas chromatography-flame ionization detection (HS-GC-FID), and inductive coupled plasma-mass spectroscopy (ICP-MS)
  • Identification of 24 non-psychoactive cannabinoids in Cannabis Sativa extracts by liquid chromatography-UV detector and using linear retention index (LRI) approach

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E C Kearse et al.
Current eye research, 20(6), 496-501 (2000-09-12)
To determine the transcorneal flux, and intracorneal penetration, of Delta9-tetrahydrocannabinol when presented to the isolated rabbit cornea in different vehicles. Corneas were mounted in specular microscope chambers, with ( 3)H-Delta9-tetrahydrocannabinol on the epithelial surface in one of 15 vehicles and
David M Wilson et al.
Drug and alcohol dependence, 67(3), 259-267 (2002-07-20)
The goal of the present study was to formulate a Delta(9)-tetrahydrocannabinol (Delta(9)-THC) metered-dose inhaler (MDI) that can be used to provide a systemic dose of Delta(9)-THC via inhalation. Following physiochemical characterization and accelerated stability testing of the aerosol, mice were
D J Harvey et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 96(1), 65-69 (1990-01-01)
1. Liver microsomes were prepared from rats, rabbits, guinea pigs, hamsters, gerbils, a cat and three strains of mice, and were incubated with delta-11-tetrahydrocannabinol (delta-11-THC). The extracted metabolites were separated by chromatography on Sephadex LH-20 and examined by gas chromatography
G Carta et al.
Brain research, 809(1), 1-4 (1998-10-31)
The effects of acute and repeated administration of Delta9-tetrahydrocannabinol (Delta9-THC), the psychoactive principle of marijuana, on acetylcholine release in the hippocampus was studied in freely moving rats by microdialysis. The acute intraperitoneal (i.p.) administration of Delta9-THC at the doses of
P M Beardsley et al.
The Journal of pharmacology and experimental therapeutics, 241(2), 521-526 (1987-05-01)
The present studies examine some of the pharmacological effects of delta-9 (11)-tetrahydrocannabinol (delta 9-11-THC), an analog of delta-9-tetrahydrocannabinol (delta 9-THC). In tests with mice, delta 9-11-THC was similar to but less potent than delta 9-THC in producing hypothermia, analgesia, lethality

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service