T-093
Delta9-Tetrahydrocannabinolic acid A solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
THCA-A
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in acetonitrile
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
cannabis testing
cannabis testing
format
single component solution
shipped in
dry ice
storage temp.
−70°C
SMILES string
OC1=C(C(O)=O)C(CCCCC)=CC2=C1[C@H](C=C(C)CC3)[C@@H]3C(C)(C)O2
InChI
1S/C22H30O4/c1-5-6-7-8-14-12-17-19(20(23)18(14)21(24)25)15-11-13(2)9-10-16(15)22(3,4)26-17/h11-12,15-16,23H,5-10H2,1-4H3,(H,24,25)/t15-,16-/m1/s1
InChI key
UCONUSSAWGCZMV-HZPDHXFCSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Δ9-Tetrahydrocannabinolic acid A (THCA-A) is a non-psychoactive acidic cannabinoid found in Cannabis that reportedly exhibits anti-inflammatory, neuroprotective, and anti-cancer effects. It is also the precursor to the main psychoactive cannabinoid found in cannabis, called Δ-9-tetrahydrocannabinol (Δ-9-THC).
Application
- Gas chromatographic-mass spectrometric (GC-MS) analysis of ten main cannabinoids from hemp inflorescences following their silylation and esterification for derivatization
- Development and validation of an HPLC-DAD method for the simultaneous analysis of five major neutral and acidic cannabinoids in cannabis and hashish samples
- Multi-analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
- Analysis of hemp seed oil samples for identification and estimation of seven major cannabinoids by HPLC coupled to a UV detector
- Estimation of ∆9-tetrahydrocannabinolic acid in blood and plasma samples using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
- Simultaneous analysis of ∆9-tetrahydrocannabinol, ∆9-tetrahydrocannabinolic acid-A, cannabidiol, and cannabinol by an HPLC-DAD method in confiscated cannabis products
Features and Benefits
- Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
- Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
- Rigorously tested through real-time stability studies to ensure accuracy and shelf life
- Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
- Flame sealed under argon into ampoules for long-term shelf life
- Offered in a convenient, DEA-exempt format to improve laboratory efficiency
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service