Skip to Content
Merck
All Photos(1)

Key Documents

C-153

Supelco

Cannabinolic Acid (CBNA) solution

1.0 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O4
CAS Number:
Molecular Weight:
354.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

shipped in

dry ice

storage temp.

−70°C

SMILES string

O1C(c2c(cc(cc2)C)c3c1cc(c(c3O)C(=O)O)CCCCC)(C)C

InChI

1S/C22H26O4/c1-5-6-7-8-14-12-17-19(20(23)18(14)21(24)25)15-11-13(2)9-10-16(15)22(3,4)26-17/h9-12,23H,5-8H2,1-4H3,(H,24,25)

InChI key

KXKOBIRSQLNUPS-UHFFFAOYSA-N

General description

CThis Certified Spiking Solution® is suitable for cannabinolic acid testing methods by GC/MS, LC/MS or HPLC for applications in the testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Cannabinolic Acid is a non-psychoactive phytocannabinoid formed upon the oxidation of Δ9-THCA in Cannabis.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Simultaneous determination of 16 phytocannabinoids in 27 samples of seized cannabis and 9 medicines based on cannabis by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Quantitative analysis of 17 cannabinoids in cannabis and hemp samples by LC-MS/MS following their liquid-solid extraction following AOAC and ASTM guidelines for cannabis and hemp matrices
  • Liquid chromatographic (LC)-diode array detection (DAD) based separation and measurement of 20 cannabinoids from a variety of cannabis samples
  • Serum analysis to identify and quantify 18 phytocannabinoids by liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathieu L Viger et al.
Journal of controlled release : official journal of the Controlled Release Society, 171(3), 308-314 (2013-06-25)
Though accurately evaluating the kinetics of release is critical for validating newly designed therapeutic carriers for in vivo applications, few methods yet exist for release measurement in real time and without the need for any sample preparation. Many of the
Awang Hazmi Awang-Junaidi et al.
Journal of animal science and biotechnology, 9, 8-8 (2018-01-27)
Gonocytes give rise to spermatogonial stem cells, and thereby play an essential role in establishing spermatogenesis. Optimized culture conditions for gonocytes provide an opportunity for their study and in vitro manipulation for potential application in reproductive technologies. Using six experiments
Tat-Chuan Cham et al.
Cells, 10(7) (2021-08-08)
The creation of a testis organoid (artificial testis tissue) with sufficient resemblance to the complex form and function of the innate testis remains challenging, especially using non-rodent donor cells. Here, we report the generation of an organoid culture system with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service