C-141
Cannabigerol solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
CBG solution
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
cannabis testing
cannabis testing
format
single component solution
storage temp.
−20°C
SMILES string
CCCCCC1=CC(O)=C(C/C=C(C)/CCC=C(C)C)C(O)=C1
InChI
1S/C21H32O2/c1-5-6-7-11-18-14-20(22)19(21(23)15-18)13-12-17(4)10-8-9-16(2)3/h9,12,14-15,22-23H,5-8,10-11,13H2,1-4H3/b17-12+
InChI key
QXACEHWTBCFNSA-SFQUDFHCSA-N
General description
Cannabigerol is a non-psychotropic phytocannabinoid that belongs to the group of C21 terpenophenolic compounds.
Application
- High-pressure liquid chromatography (HPLC)- diode array detector (DAD) based quantification of cannabinoids in cannabis plant material
- Development and validation of an HPLC method to identify and quantify 11 cannabinoids in different varieties of cannabis plant materials
- Determination of eight cannabinoids in recreational and forensic hemp samples using reversed phase-high-pressure liquid chromatography (RP-HPLC) in combination with UV detector
- Extraction of nine cannabinoids from food and beverage samples having hemp seeds, oil, and flour as ingredients followed by their analysis by liquid chromatography-electrospray ionization-QTrap-tandem mass spectrometry LC/ESI‐QTRAP‐MS/MS
- High-pressure liquid chromatography (HPLC)-UV detection-based determination of main cannabinoids in 13 commercially available hemp seed oil samples
- Simultaneous analysis of five major cannabinoids and their metabolites in human urine and plasma samples by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS)
Features and Benefits
- Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
- Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
- Rigorously tested through real-time stability studies to ensure accuracy and shelf life
- Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
- Flame sealed under argon into ampoules for long-term shelf life
- Offered in a convenient, DEA-exempt format to improve laboratory efficiency
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service