Skip to Content
Merck
All Photos(1)

Key Documents

T-003

Supelco

(−)-Δ9-THC-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27O2D3
CAS Number:
Molecular Weight:
317.48
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

−20°C

SMILES string

O1C(C2C(c3c1cc(cc3O)CCCCC([2H])([2H])[2H])C=C(CC2)C)(C)C

InChI

1S/C21H30O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h11-13,16-17,22H,5-10H2,1-4H3/i1D3

InChI key

CYQFCXCEBYINGO-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

It is an internal standard of Δ9-THC, the main active constituent and the primary psychoactive cannabinoid of cannabis.

Application

This Certified Spiking Solution® is suitable as an internal standard for quantitation of Δ9-THC levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. The Certified Reference Material (CRM) in solution can also be used as follows:
  • Development of an automated sample preparation method based on solid phase extraction (SPE) to determine ∆9-tetrahydrocannabinol (THC) and its three primary metabolites in a variety of matrices, such as serum, saliva, and urine using liquid chromatography-mass spectrometry (LC-MS)
  • Comparative analysis of two methods based on ultra high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method and solid phase microextraction-gas chromatography-mass spectrometry (SPME-GC/MS) to measure cannabinoids in oral fluid samples of small volumes
  • As an internal standard in high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) based quantitative assay of 11 cannabinoids and their metabolites in human urine and plasma samples

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueyao Zhang et al.
PloS one, 9(12), e114776-e114776 (2014-12-09)
Glutathione S-transferases (GSTs) are multifunctional enzymes that are involved in the metabolism of endogenous and exogenous compounds and are related to insecticide resistance. The purpose of this study was to provide new information on the molecular characteristics and the positive
Alice Palermiti et al.
Pharmaceuticals (Basel, Switzerland), 14(5) (2021-06-03)
Medical cannabis is increasingly being used in the treatment and support of several diseases and syndromes. The quantitative determination of active ingredients (delta-9 tetrahydrocannabinol, THC, and cannabidiol, CBD) in galenic oily preparations is prescribed by law for each produced batch.
Federica Pigliasco et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-14)
The aim of this work is to evaluate volumetric absorptive microsampling (VAMS) from capillary blood as an alternative strategy for therapeutic drug monitoring (TDM) in patients treated with the newly available GW-purified form of cannabidiol (Epidiolex®). A fast ultra-high-performance liquid
Luisa A Wetta et al.
American journal of perinatology, 31(6), 541-546 (2013-09-12)
The objective of this study is to evaluate whether midtrimester maternal vitamin D is associated with preeclampsia < 37 weeks or spontaneous preterm birth (SPTB) < 35 weeks. Nested case-control comprising two case subsets: (1) 100 women with preeclampsia < 37 weeks and (2) 100 women

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service