Skip to Content
Merck
All Photos(1)

Key Documents

T-018

Supelco

(−)-11-nor-9-Carboxy-Δ9-THC solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

suitability

suitable for immunoassay

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCCCCC(C=C1O)=CC2=C1[C@@]3([H])[C@@](C(C)(C)O2)([H])CCC(C(O)=O)=C3

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/t15-,16-/m1/s1

InChI key

YOVRGSHRZRJTLZ-HZPDHXFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An analytical reference standard suitable in cannabinoid testing, clinical toxicology, forensic analysis, or urine drug testing by LC-MS/MS or GC/MS. Also known as THC acid, 11-nor-9-Carboxy-Δ9-THC is the main secondary metabolite of THC found in urine or blood of cannabis users.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Minoli et al.
Forensic science international, 218(1-3), 49-52 (2011-11-01)
The fine detection of 11-nor-Δ9-tetrahydrocannabinol-9-carboxylic acid (THCCOOH) in hair matrix remains one of the most important topics in hair analysis. This relevance lies in the necessity to obtain evidence of effective drug consumption and dispel any doubt of environmental contamination.
Xiang He et al.
Analytical chemistry, 84(18), 7643-7647 (2012-08-21)
Oral fluid has been gaining more acceptance as the alternative matrix for forensic toxicology. Currently, Δ(9)-tetrahydrocannabinol (THC) is used as the primary target for detecting cannabis use in oral fluid. Meanwhile, THC carboxylic acid (THCA) in oral fluid is reported
Cynthia Coulter et al.
Journal of analytical toxicology, 36(6), 413-417 (2012-04-26)
This paper describes the determination of tetrahydrocannabinol (THC) and its metabolite, 11-nor-Δ⁹-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) in oral fluid using solid-phase extraction and liquid chromatography with tandem mass spectral detection (LC-MS-MS) and its application to proficiency specimens. The method employs collection of
Vanessa de M Prata et al.
Analytical and bioanalytical chemistry, 403(2), 625-632 (2012-03-01)
A new procedure is described for the derivatization by silylation of 11-nor-Δ(9)-tetrahydrocannabinol-9-carboxylic acid (THCCOOH) present in urine, followed by analysis using gas chromatography-tandem mass spectrometry. A conventional procedure for derivatization of the analyte was evaluated using two types of experimental
Yannick Lecompte et al.
Journal of analytical toxicology, 36(8), 569-574 (2012-08-31)
The guidelines for screening of urinary cannabinoids require that all specimens testing positive should be confirmed by gas chromatography-mass spectrometry at a confirmatory test cutoff value of 15 ng/mL of 11-nor-9-carboxy-Δ9-tetrahydrocannabinol (THCCOOH). To assess the impact of lowering the confirmatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service