Skip to Content
Merck
All Photos(2)

Documents

T62405

Sigma-Aldrich

Silver trifluoroacetate

98%

Synonym(s):

Trifluoroacetic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOAg
CAS Number:
Molecular Weight:
220.88
Beilstein:
3634022
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

reaction suitability

core: silver
reagent type: catalyst

mp

257-260 °C (dec.) (lit.)

SMILES string

FC(F)(F)C(=O)O[Ag]

InChI

1S/C2HF3O2.Ag/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

KZJPVUDYAMEDRM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Silver trifluoroacetate can be used as a precursor tosynthesize silver nanoparticles for various applications. For example, it isused to prepare Pt-on-Ag bimetallic heteronanostructures and nanofibrousbiosensors.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helvetica Chimica Acta, 72, 1362-1362 (1989)
Teodora Miclăuş et al.
Nano letters, 14(4), 2086-2093 (2014-03-13)
Protein coronas around silver nanocubes were quantified in serum-containing media using localized surface plasmon resonances. Both soft and hard coronas showed exposure-time and concentration-dependent changes in protein surface density with time-dependent hardening. We observed spatially dependent kinetics of the corona-formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service