![Silver trifluoroacetate 98%](/deepweb/assets/sigmaaldrich/product/structures/265/936/36978a40-a68c-4e6c-bdc1-15d952655ac0/640/36978a40-a68c-4e6c-bdc1-15d952655ac0.png)
Silver trifluoroacetate
![Silver trifluoroacetate 98%](/deepweb/assets/sigmaaldrich/product/structures/265/936/36978a40-a68c-4e6c-bdc1-15d952655ac0/640/36978a40-a68c-4e6c-bdc1-15d952655ac0.png)
Synonym(s):
Trifluoroacetic acid silver salt, Silver trifluoroacetate
Linear Formula:
CF3COOAg
CAS Number:
Molecular Weight:
220.88
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-3 of 3
Compare
Product Number
Description
Pricing