Skip to Content
Merck
All Photos(2)

Key Documents

208361

Sigma-Aldrich

Silver tetrafluoroborate

98%

Synonym(s):

Silver borofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgBF4
CAS Number:
Molecular Weight:
194.67
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: silver
reagent type: catalyst

mp

70-73 °C (lit.)

SMILES string

[Ag+].F[B-](F)(F)F

InChI

1S/Ag.BF4/c;2-1(3,4)5/q+1;-1

InChI key

CCAVYRRHZLAMDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver tetrafluoroborate (AgBF4) is used as a Lewis acid in organic synthesis. It is known to have a high affinity towards organic halides. It is employed in organic reactions like activation of acyl chlorides, nucleophilic substitution, rearrangement, cycloaddition, cyclization, ring expansion and oxidation.

Application

Displaces chloride from precious metal complexes used in the oxidation of alcohols by persulfate.
Provides a silver-ion template for improved macrolatamization of a linear dipeptide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mol. Catal., 79, 85-85 (1993)
Kende, A. S.; Huang, H.
Tetrahedron Letters, 38, 3353-3353 (1997)
Silver tetrafluoroborate
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2005)
Synlett, 1901-1901 (2007)
Silver tetrafluoroborate
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service