Skip to Content
Merck
All Photos(2)

Key Documents

679658

Sigma-Aldrich

Tris(4-formylphenyl)amine

97%

Synonym(s):

4,4′,4″-Nitrilotrisbenzaldehyde, 4,4′,4″-Triformyltriphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15NO3
CAS Number:
Molecular Weight:
329.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

244-248 °C

SMILES string

[H]C(=O)c1ccc(cc1)N(c2ccc(cc2)C([H])=O)c3ccc(cc3)C([H])=O

InChI

1S/C21H15NO3/c23-13-16-1-7-19(8-2-16)22(20-9-3-17(14-24)4-10-20)21-11-5-18(15-25)6-12-21/h1-15H

InChI key

YOXHQRNDWBRUOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in preparation of triangular ligands for self-assembly into M4L4 tetrahedra.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3736-3736 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service