Skip to Content
Merck
All Photos(2)

Documents

483346

Sigma-Aldrich

Silver trifluoromethanesulfonate

≥99.95% trace metals basis

Synonym(s):

AgOTf, Silver triflate, Trifluoromethanesulfonic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Ag
CAS Number:
Molecular Weight:
256.94
Beilstein:
3598402
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.95% trace metals basis

reaction suitability

core: silver
reagent type: catalyst

mp

286 °C (lit.)

SMILES string

[Ag+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Ag/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

QRUBYZBWAOOHSV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Silver trifluoromethanesulfonate (AgOTf ) is a reactive triflating agent, which converts alkyl, acyl and sulfonyl halides to corresponding triflate species.
  • It is a highly suitable electrophile to initiate acetylenic oxy-Cope rearrangement of substituted 5-hexen-1-yn-3-ols to synthesize corresponding α,δ-diethylenic aldehydes.
  • It can also be used in the diastereoselective cyclization of amino ketenes where the diastereoselectivity depends on Ag(I) concentration.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver mediated acetylenic oxy cope rearrangement.
Bluthe N, et al.
Tetrahedron, 42(5), 1333-1344 (1986)
Asymmetric synthesis via electrophile-mediated cyclisations.
Fox D N and Gallagher T
Tetrahedron, 46(13-14), 4697-4710 (1990)
Trifluoromethanesulfonic?Carboxylic Anhydrides, Highly Active Acylating Agents.
Effenberger F and Epple G
Angewandte Chemie (International Edition in English), 11(4), 299-300 (1972)
Perfluoroalkanesulfonic esters: methods of preparation and applications in organic chemistry.
Stang P J, et al.
Synthesis, 1982(02), 85-126 (1982)
Yusuke Imazaki et al.
Journal of the American Chemical Society, 134(36), 14760-14763 (2012-08-25)
Aryl triflates were transformed to aryl bromides/iodides simply by treating them with LiBr/NaI and [Cp*Ru(MeCN)(3)]OTf. The ruthenium complex also catalyzed the transformation of alkenyl sulfonates and phosphates to alkenyl halides under mild conditions. Aryl and alkenyl triflates undergo oxidative addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service