Skip to Content
Merck
All Photos(4)

Documents

176435

Sigma-Aldrich

Silver trifluoromethanesulfonate

≥99%

Synonym(s):

Silver triflate, Trifluoromethanesulfonic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Ag
CAS Number:
Molecular Weight:
256.94
Beilstein:
3598402
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

powder

reaction suitability

core: silver
reagent type: catalyst

mp

286 °C (lit.)

SMILES string

[Ag+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Ag/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

QRUBYZBWAOOHSV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Silver trifluoromethanesulfonate is a reactive triflating agent and source of soluble silver ions useful for formation of electrophilic onium species, and promotion of Friedel-Crafts, nucleophile alkene cyclization, and esterification reactions.
It can also be used:
  • To obtain olefins from secondary phosphates and thiophosphates.
  • As a reagent in the etherification of alcohols with primary alkyl halides under mild conditions.
  • To generate cationic rhodium catalysts from chlororhodium complexes for the hydrophosphination of acetylenes.
  • As a catalyst for the preparation of silyl ethers by hydrosilylation of aldehydes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Communications (Cambridge, England), 1359-1359 (2006)
New synthetic methods for olefins from secondary phosphates and thiophosphates
Shimagaki M, et al.
Tetrahedron Letters, 36, 719-719 (1995)
Minoru Hayashi et al.
The Journal of organic chemistry, 71(24), 9248-9251 (2006-11-18)
A novel rhodium-catalyzed hydrophosphination of alkynes using a silylphosphine as a phosphino group source is described. A variety of alkynes, both terminal and internal ones with aryl, alkyl, and carboxyl groups, gave the corresponding alkenylphosphines in a highly regioselective and
A mild procedure for etherification of alcohols with primary alkyl halides in the presence of silver triflate
Burk RM, et al.
Tetrahedron Letters, 35, 8111-8111 (1994)
Simbarashe Ngwerume et al.
The Journal of organic chemistry, 78(3), 920-934 (2012-12-29)
A novel gold-catalyzed method for the regioselective synthesis of highly substituted pyrroles directly from oximes and alkynes was developed via independent optimization of two key steps of the process. Importantly, a cationic gold(I) species was shown to activate multiple steps

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service