Skip to Content
Merck
All Photos(3)

Documents

K0875

Sigma-Aldrich

Sodium 2-oxobutyrate

powder

Synonym(s):

2-Oxobutanoic acid sodium salt, 2-Oxobutyric acid sodium salt, Sodium α-ketobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOONa
CAS Number:
Molecular Weight:
124.07
Beilstein:
3631701
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

color

white

storage temp.

2-8°C

SMILES string

[Na+].CCC(=O)C([O-])=O

InChI

1S/C4H6O3.Na/c1-2-3(5)4(6)7;/h2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

SUAMAHKUSIHRMR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium 2-oxobutyrate can be used in the preparation of metal complexes such as lanthanide poly(imino carboxylate) complexes and half-sandwich complexes of (S)-1-amino-2-(methoxymethyl)-pyrrolidine. It can also be used in the synthesis of antiviral agents, 6-azapyrimidine-2′-deoxy-4′-thionucleosides.
Substrate for the determination of lactate dehydrogenase isoenzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

6-Azapyrimidine-2`-deoxy-4`-thionucleosides: Antiviral Agents against TK+ and TK? HSV and VZV Strains.
Maslen H L, et al.
Journal of Medicinal Chemistry, 47(22), 5482-5491 (2004)
Formation of oligomeric lanthanide complexes with new tripodal poly (imino carboxylate) ligands.
Blake A J, et al.
J. Chem. Soc., Dalton Trans., 20, 3655-3658 (1997)
Half-sandwich complexes of (S)-1-amino-2-(methoxymethyl)-pyrrolidine (SAMP).
Hoffmuller W, et al.
Journal of Organometallic Chemistry, 564(1), 179-187 (1998)
Shufen Huang et al.
Journal of molecular biology, 396(3), 708-718 (2009-12-08)
In recent years, increased interest has been directed towards hydrogen sulfide (H2S) as the third gasotransmitter and its role in various diseases. Cystathionine-gamma-lyase (CSE) is one of the enzymes responsible for the endogenous production of H2S in mammals. With the
Sergey V Smirnov et al.
FEMS microbiology letters, 273(1), 70-77 (2007-06-15)
A two-step enzymatic synthesis process of 4-hydroxyisoleucine is suggested. In the first step, the aldol condensation of acetaldehyde and alpha-ketobutyrate catalyzed by specific aldolase results in the formation of 4-hydroxy-3-methyl-2-keto-pentanoate (HMKP). In the second step, amination of HMKP by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service