Skip to Content
Merck
All Photos(3)

Documents

K0629

Sigma-Aldrich

Sodium 4-methyl-2-oxovalerate

leucine metabolite

Synonym(s):

α-Ketoisocaproic acid sodium salt, 4-Methyl-2-oxopentanoic acid sodium salt, 4-Methyl-2-oxovaleric acid sodium salt, Ketoleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCOONa
CAS Number:
Molecular Weight:
152.12
Beilstein:
4239297
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

Assay

>98% (TLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

mp

275 °C (dec.) (lit.)

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

[Na+].CC(C)CC(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-4(2)3-5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1

InChI key

IXFAZKRLPPMQEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 4-methyl-2-oxovalerate has been used for quantifying branched-chain keto acids from cell extracts by HPLC-fluorescence method.

Biochem/physiol Actions

α-Ketoisocaproic acid is a leucine metabolite that is known to induce insulin secretion from the βcells of pancreas. Enteral infusion of aα-Ketoisocaproate is found to ameliorate, endotoxemic condition. The resulting ketone bodies obtained from its conversion, might serve as an energy source. Accumulation of α-Ketoisocaproic acid is characterized in branched chain ketoaciduria, which is caused due to the lack of branched chain L-2-keto acid dehydrogenase activity. α-Ketoisocaproic acid causes dissociation in the oxidative phosphorylation reaction and acts as a metabolic inhibitor of α-ketoglutarate dehydrogenase. Thus, leads to a defect in the mitochondrial homeostasis, observed in branched chain ketoaciduria.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enteral infusion of sodium 2-ketoisocaproate in endotoxic rats.
Hirokawa M and Walser M
Critical Care Medicine, 27(2), 373-379 (1999)
Effects of L-leucine and alpha-ketoisocaproic acid upon insulin secretion and metabolism of isolated pancreatic islets.
Panten U
Febs Letters, 20(2), 225-228 (1972)
Branched chain a- Keto acid metabolism
JOE A. BOWDEN
The Journal of Biological Chemistry, 243, 3526-3531 (1968)
Analysis of Branched-Chain Keto Acids in Cell Extracts by HPLC-Fluorescence Detention
Ayuna HATTORI
Chromatography, 38, 129-133 (2017)
Yuhong Zhu et al.
Enzyme and microbial technology, 49(4), 321-325 (2011-11-25)
This work aims to produce α-ketoisocaproate (KIC) from L-leucine via the free-whole-cell biotransformation of Rhodococcus opacus DSM 43250. The effects of temperature, pH, substrate concentration, cell concentration, and rotating speed on KIC production were examined. Furthermore, the biotransformation conditions were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service