Skip to Content
Merck
All Photos(6)

Documents

198994

Sigma-Aldrich

Sodium 3-methyl-2-oxobutyrate

greener alternative

95%

Synonym(s):

α-Ketoisovaleric acid sodium salt, 2-Keto-3-methylbutyric acid sodium salt, 3-Methyl-2-oxobutanoic acid sodium salt, 3-Methyl-2-oxobutyric acid sodium salt, Ketovaline sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCOONa
CAS Number:
Molecular Weight:
138.10
Beilstein:
4334552
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

greener alternative product score

old score: 14
new score: 9
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

220-230 °C (dec.) (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

greener alternative category

SMILES string

[Na+].CC(C)C(=O)C([O-])=O

InChI

1S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1

InChI key

WIQBZDCJCRFGKA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 3-methyl-2-oxobutyrate (3-Methyl-2-oxobutanoic acid sodium salt) was used in the synthesis of (S)-2-hydroxy-3-methylbutanoic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An evaluation of the substrate specificity and asymmetric synthesis potential of the cloned L-lactate dehydrogenase from Bacillus stearothermophilus.
Canadian Journal of Chemistry, 67(6), 1065-1070 (1989)
Marta de la Plaza et al.
Journal of molecular microbiology and biotechnology, 17(2), 96-100 (2008-11-27)
The alpha-ketoisovalerate decarboxylase (Kivd) is a unique lactococcal key enzyme in the decarboxylation of branched-chain alpha-keto acids derived from branched-chain amino acids transamination into aldehydes. These products are important aroma compounds produced during cheese ripening. In this study, the Kivd
Taro Murakami et al.
Journal of nutritional science and vitaminology, 51(1), 48-50 (2005-05-27)
Branched-chain alpha-keto acid dehydrogenase kinase (BDK) phosphorylates and inactivates the branched-chain alpha-keto acid dehydrogenase complex (BCKDC), which is the rate-limiting enzyme in the branched-chain amino acid catabolism. BDK has been believed to be bound to the BCKDC. However, recent our
Joachim Schuster et al.
Plant molecular biology, 57(2), 241-254 (2005-04-12)
Plants are capable to de novo synthesize the essential amino acids leucine, isoleucine and valine. Studies in recent years, however, also revealed that plants have the potential to degrade leucine or may be all of the branched-chain amino acids. One
Cláudia Funchal et al.
Metabolic brain disease, 20(3), 205-217 (2005-09-17)
In this study we investigate the effects of the branched-chain keto acids (BCKA) alpha-ketoisocaproic (KIC), alpha-ketoisovaleric (KIV), and alpha-keto-beta-methylvaleric (KMV) acids, metabolites accumulating in maple syrup urine disease (MSUD), on the in vitro phosphorylation of glial fibrillary acidic protein (GFAP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service